Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69276994-69277237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC125
Entrez Gene ID
202243
Gene Name
coiled-coil domain containing 125
Gene Aliases
KENAE
UniGene
Hs.654850
Gene Chromosome Location
Chr.5: 69273087-69333069 on Build GRCh38
Transcripts
XM_011543258.2 NM_001324069.1 NM_001324070.1 NM_001324071.1 NM_001324072.1 NM_001324074.1 NM_001324075.1 NM_001324077.1 NM_001324078.1 NM_001799.3 NR_136690.1 XM_011543094.2

Gene Symbol
CDK7
Entrez Gene ID
1022
Gene Name
cyclin dependent kinase 7
Gene Aliases
CAK, CAK1, CDKN7, HCAK, MO15, STK1, p39MO15
UniGene
Hs.184298
Gene Chromosome Location
Chr.5: 69234795-69277430 on Build GRCh38
Transcripts
XM_011543258.2 NM_001324069.1 NM_001324070.1 NM_001324071.1 NM_001324072.1 NM_001324074.1 NM_001324075.1 NM_001324077.1 NM_001324078.1 NM_001799.3 NR_136690.1 XM_011543094.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069712 c.1032A>G p.L344L Chr.5: 69277126-69277126 on Build GRCh38
4671184 c.1026G>T p.K342N Chr.5: 69277120-69277120 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767939604 Chr.5: 69277075-69277076 on Build GRCh38
rs1009315224 Chr.5: 69277142-69277142 on Build GRCh38
rs781234036 Chr.5: 69277104-69277104 on Build GRCh38
rs572455178 Chr.5: 69277049-69277049 on Build GRCh38
rs772450221 Chr.5: 69277162-69277162 on Build GRCh38
rs745819872 Chr.5: 69277112-69277112 on Build GRCh38
rs769162123 Chr.5: 69277117-69277117 on Build GRCh38
rs774000048 Chr.5: 69277152-69277152 on Build GRCh38
rs1047847744 Chr.5: 69277043-69277043 on Build GRCh38
rs1053622317 Chr.5: 69277110-69277110 on Build GRCh38
rs545813153 Chr.5: 69277151-69277151 on Build GRCh38
rs748475019 Chr.5: 69277154-69277154 on Build GRCh38
rs777416496 Chr.5: 69277081-69277081 on Build GRCh38
rs774469602 Chr.5: 69277076-69277076 on Build GRCh38
rs751340229 Chr.5: 69277085-69277085 on Build GRCh38
rs373823744 Chr.5: 69277066-69277066 on Build GRCh38
rs911953518 Chr.5: 69277047-69277047 on Build GRCh38
rs199570737 Chr.5: 69277085-69277086 on Build GRCh38
rs757120567 Chr.5: 69277089-69277089 on Build GRCh38
rs748884249 Chr.5: 69277144-69277144 on Build GRCh38
rs753976030 Chr.5: 69277056-69277056 on Build GRCh38
rs779651538 Chr.5: 69277127-69277127 on Build GRCh38
rs750011856 Chr.5: 69277137-69277137 on Build GRCh38
rs1043905884 Chr.5: 69277156-69277156 on Build GRCh38
rs767542729 Chr.5: 69277127-69277128 on Build GRCh38
rs1020674278 Chr.5: 69277194-69277194 on Build GRCh38

View Full Product Details