Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114718062-114718311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSDE1
Entrez Gene ID
7812
Gene Name
cold shock domain containing E1
Gene Aliases
D1S155E, UNR
UniGene
Hs.69855
Gene Chromosome Location
Chr.1: 114716913-114758050 on Build GRCh38
Transcripts
NM_001007553.2 NM_001130523.2 NM_001242891.1 NM_001242892.1 NM_001242893.1 NM_007158.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4784031 c.2419C>T p.R807C Chr.1: 114718192-114718192 on Build GRCh38
7001334 c.2384_2385insT p.V796fs*3 Chr.1: 114718181-114718182 on Build GRCh38
6835247 c.2401G>C p.G801R Chr.1: 114718210-114718210 on Build GRCh38
894242 c.2375G>A p.R792H Chr.1: 114718191-114718191 on Build GRCh38
4868320 c.2420G>A p.R807H Chr.1: 114718191-114718191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774579235 Chr.1: 114718209-114718209 on Build GRCh38
rs147036044 Chr.1: 114718256-114718256 on Build GRCh38
rs770432178 Chr.1: 114718121-114718121 on Build GRCh38
rs908473995 Chr.1: 114718107-114718107 on Build GRCh38
rs531344093 Chr.1: 114718214-114718214 on Build GRCh38
rs750274957 Chr.1: 114718118-114718118 on Build GRCh38
rs375339756 Chr.1: 114718191-114718191 on Build GRCh38
rs749461339 Chr.1: 114718266-114718266 on Build GRCh38
rs1036478630 Chr.1: 114718244-114718244 on Build GRCh38
rs546258811 Chr.1: 114718243-114718243 on Build GRCh38
rs773967564 Chr.1: 114718241-114718241 on Build GRCh38
rs369948708 Chr.1: 114718159-114718159 on Build GRCh38
rs753260110 Chr.1: 114718136-114718136 on Build GRCh38
rs559081952 Chr.1: 114718229-114718230 on Build GRCh38
rs9724612 Chr.1: 114718160-114718160 on Build GRCh38
rs372889272 Chr.1: 114718264-114718266 on Build GRCh38
rs777289026 Chr.1: 114718137-114718137 on Build GRCh38
rs183844328 Chr.1: 114718157-114718157 on Build GRCh38
rs778572647 Chr.1: 114718220-114718221 on Build GRCh38
rs373035712 Chr.1: 114718245-114718245 on Build GRCh38
rs764762466 Chr.1: 114718240-114718240 on Build GRCh38
rs759111720 Chr.1: 114718239-114718239 on Build GRCh38
rs769874747 Chr.1: 114718180-114718180 on Build GRCh38
rs765946586 Chr.1: 114718225-114718225 on Build GRCh38
rs766109129 Chr.1: 114718122-114718122 on Build GRCh38
rs753588418 Chr.1: 114718230-114718230 on Build GRCh38
rs761802148 Chr.1: 114718220-114718220 on Build GRCh38
rs779486463 Chr.1: 114718170-114718170 on Build GRCh38
rs760562179 Chr.1: 114718223-114718223 on Build GRCh38
rs1047140469 Chr.1: 114718098-114718098 on Build GRCh38
rs1019816797 Chr.1: 114718276-114718276 on Build GRCh38
rs768724629 Chr.1: 114718192-114718192 on Build GRCh38
rs1801909 Chr.1: 114718141-114718141 on Build GRCh38
rs150575144 Chr.1: 114718188-114718188 on Build GRCh38
rs745746242 Chr.1: 114718228-114718229 on Build GRCh38
rs553300119 Chr.1: 114718100-114718100 on Build GRCh38
rs775636582 Chr.1: 114718230-114718230 on Build GRCh38
rs11558869 Chr.1: 114718207-114718207 on Build GRCh38
rs780294044 Chr.1: 114718262-114718262 on Build GRCh38
rs373120787 Chr.1: 114718261-114718261 on Build GRCh38
rs746279844 Chr.1: 114718176-114718176 on Build GRCh38
rs755778971 Chr.1: 114718121-114718121 on Build GRCh38
rs374095550 Chr.1: 114718168-114718168 on Build GRCh38
rs112218640 Chr.1: 114718260-114718260 on Build GRCh38
rs9724613 Chr.1: 114718117-114718117 on Build GRCh38
rs753210209 Chr.1: 114718270-114718270 on Build GRCh38
rs1045973072 Chr.1: 114718274-114718274 on Build GRCh38
rs189058114 Chr.1: 114718221-114718221 on Build GRCh38

View Full Product Details