Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53636565-53636836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPRX
Entrez Gene ID
503834
Gene Name
divergent-paired related homeobox
Gene Aliases
-
UniGene
Hs.579941
Gene Chromosome Location
Chr.19: 53601257-53637057 on Build GRCh38
Transcripts
NM_001012728.1 XM_011527011.2 XM_011527012.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319996 c.424A>G p.N142D Chr.19: 53636836-53636836 on Build GRCh38
6649054 c.348G>A p.P116P Chr.19: 53636760-53636760 on Build GRCh38
4570106 c.234T>C p.H78H Chr.19: 53636646-53636646 on Build GRCh38
6649052 c.221C>T p.A74V Chr.19: 53636633-53636633 on Build GRCh38
2928915 c.287C>T p.S96F Chr.19: 53636699-53636699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965564416 Chr.19: 53636717-53636717 on Build GRCh38
rs745918887 Chr.19: 53636791-53636791 on Build GRCh38
rs770924095 Chr.19: 53636646-53636646 on Build GRCh38
rs140829869 Chr.19: 53636709-53636709 on Build GRCh38
rs565196699 Chr.19: 53636817-53636817 on Build GRCh38
rs765836576 Chr.19: 53636687-53636687 on Build GRCh38
rs202096492 Chr.19: 53636774-53636774 on Build GRCh38
rs1009518882 Chr.19: 53636722-53636722 on Build GRCh38
rs745996785 Chr.19: 53636740-53636740 on Build GRCh38
rs769146467 Chr.19: 53636750-53636750 on Build GRCh38
rs371523408 Chr.19: 53636764-53636764 on Build GRCh38
rs779696506 Chr.19: 53636596-53636596 on Build GRCh38
rs376708716 Chr.19: 53636815-53636815 on Build GRCh38
rs768111180 Chr.19: 53636761-53636761 on Build GRCh38
rs150237904 Chr.19: 53636785-53636785 on Build GRCh38
rs777746040 Chr.19: 53636638-53636638 on Build GRCh38
rs776875397 Chr.19: 53636656-53636656 on Build GRCh38
rs1025988724 Chr.19: 53636680-53636680 on Build GRCh38
rs775961921 Chr.19: 53636667-53636667 on Build GRCh38
rs898933951 Chr.19: 53636604-53636604 on Build GRCh38
rs781253762 Chr.19: 53636790-53636790 on Build GRCh38
rs780214207 Chr.19: 53636746-53636746 on Build GRCh38
rs755739837 Chr.19: 53636590-53636590 on Build GRCh38
rs868486840 Chr.19: 53636782-53636782 on Build GRCh38
rs764685283 Chr.19: 53636671-53636671 on Build GRCh38
rs201734272 Chr.19: 53636765-53636765 on Build GRCh38
rs778553055 Chr.19: 53636617-53636617 on Build GRCh38
rs752125092 Chr.19: 53636675-53636675 on Build GRCh38
rs374419575 Chr.19: 53636730-53636730 on Build GRCh38
rs546300623 Chr.19: 53636688-53636688 on Build GRCh38
rs779698125 Chr.19: 53636713-53636713 on Build GRCh38
rs371142421 Chr.19: 53636705-53636705 on Build GRCh38
rs762206233 Chr.19: 53636760-53636760 on Build GRCh38
rs746285867 Chr.19: 53636608-53636608 on Build GRCh38
rs937824375 Chr.19: 53636814-53636814 on Build GRCh38
rs138900772 Chr.19: 53636786-53636786 on Build GRCh38
rs769091377 Chr.19: 53636816-53636816 on Build GRCh38
rs770101038 Chr.19: 53636665-53636665 on Build GRCh38
rs144673249 Chr.19: 53636747-53636747 on Build GRCh38
rs141641052 Chr.19: 53636778-53636778 on Build GRCh38
rs368484033 Chr.19: 53636741-53636741 on Build GRCh38
rs771996860 Chr.19: 53636630-53636630 on Build GRCh38
rs763901601 Chr.19: 53636779-53636779 on Build GRCh38
rs368786641 Chr.19: 53636676-53636676 on Build GRCh38
rs201290452 Chr.19: 53636795-53636795 on Build GRCh38
rs759696783 Chr.19: 53636662-53636662 on Build GRCh38
rs753438549 Chr.19: 53636690-53636690 on Build GRCh38
rs747037499 Chr.19: 53636645-53636645 on Build GRCh38
rs763473546 Chr.19: 53636669-53636669 on Build GRCh38
rs746982581 Chr.19: 53636710-53636710 on Build GRCh38
rs575979078 Chr.19: 53636748-53636748 on Build GRCh38
rs754556982 Chr.19: 53636692-53636692 on Build GRCh38
rs770186726 Chr.19: 53636654-53636654 on Build GRCh38
rs753434011 Chr.19: 53636601-53636601 on Build GRCh38
rs778694585 Chr.19: 53636699-53636699 on Build GRCh38
rs375450549 Chr.19: 53636589-53636589 on Build GRCh38
rs747980450 Chr.19: 53636618-53636618 on Build GRCh38
rs144585982 Chr.19: 53636672-53636672 on Build GRCh38
rs368343082 Chr.19: 53636759-53636759 on Build GRCh38
rs754756196 Chr.19: 53636615-53636615 on Build GRCh38
rs756232285 Chr.19: 53636792-53636792 on Build GRCh38
rs867949523 Chr.19: 53636634-53636634 on Build GRCh38
rs138559564 Chr.19: 53636763-53636763 on Build GRCh38
rs752355371 Chr.19: 53636777-53636777 on Build GRCh38
rs1032197381 Chr.19: 53636692-53636692 on Build GRCh38

View Full Product Details