Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55256353-55256622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX29
Entrez Gene ID
54505
Gene Name
DEAH-box helicase 29
Gene Aliases
DDX29
UniGene
Hs.593268
Gene Chromosome Location
Chr.5: 55256245-55307693 on Build GRCh38
Transcripts
NM_019030.2 XM_006714653.2 XM_017009588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855599 c.4082C>T p.T1361M Chr.5: 55256516-55256516 on Build GRCh38
3617044 c.4107C>T p.N1369N Chr.5: 55256491-55256491 on Build GRCh38
7119535 c.4089G>C p.L1363F Chr.5: 55256509-55256509 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570421120 Chr.5: 55256410-55256410 on Build GRCh38
rs774552703 Chr.5: 55256552-55256553 on Build GRCh38
rs369793243 Chr.5: 55256567-55256567 on Build GRCh38
rs777482659 Chr.5: 55256457-55256457 on Build GRCh38
rs755460918 Chr.5: 55256442-55256443 on Build GRCh38
rs947740245 Chr.5: 55256429-55256429 on Build GRCh38
rs1047113457 Chr.5: 55256381-55256381 on Build GRCh38
rs192390194 Chr.5: 55256451-55256451 on Build GRCh38
rs551953510 Chr.5: 55256379-55256379 on Build GRCh38
rs201030012 Chr.5: 55256515-55256515 on Build GRCh38
rs774737992 Chr.5: 55256545-55256545 on Build GRCh38
rs751318927 Chr.5: 55256577-55256577 on Build GRCh38
rs777812201 Chr.5: 55256496-55256497 on Build GRCh38
rs770420860 Chr.5: 55256442-55256442 on Build GRCh38
rs897229822 Chr.5: 55256455-55256455 on Build GRCh38
rs777599227 Chr.5: 55256568-55256568 on Build GRCh38
rs752689422 Chr.5: 55256516-55256516 on Build GRCh38
rs781292237 Chr.5: 55256490-55256490 on Build GRCh38
rs868068205 Chr.5: 55256402-55256402 on Build GRCh38
rs762111922 Chr.5: 55256546-55256546 on Build GRCh38
rs748448665 Chr.5: 55256483-55256484 on Build GRCh38
rs1011775699 Chr.5: 55256534-55256534 on Build GRCh38
rs769388049 Chr.5: 55256525-55256525 on Build GRCh38
rs781598775 Chr.5: 55256450-55256455 on Build GRCh38
rs756375789 Chr.5: 55256491-55256500 on Build GRCh38
rs752214385 Chr.5: 55256562-55256562 on Build GRCh38
rs752700888 Chr.5: 55256440-55256440 on Build GRCh38
rs1037150560 Chr.5: 55256432-55256432 on Build GRCh38
rs757050206 Chr.5: 55256580-55256580 on Build GRCh38
rs528202866 Chr.5: 55256418-55256418 on Build GRCh38
rs568096465 Chr.5: 55256477-55256477 on Build GRCh38
rs76296338 Chr.5: 55256588-55256588 on Build GRCh38
rs769291732 Chr.5: 55256532-55256532 on Build GRCh38
rs111840507 Chr.5: 55256535-55256535 on Build GRCh38
rs749420136 Chr.5: 55256500-55256500 on Build GRCh38
rs745937824 Chr.5: 55256501-55256501 on Build GRCh38
rs745858959 Chr.5: 55256553-55256554 on Build GRCh38
rs1020030864 Chr.5: 55256551-55256551 on Build GRCh38
rs369600455 Chr.5: 55256473-55256473 on Build GRCh38
rs763497858 Chr.5: 55256528-55256528 on Build GRCh38
rs775238784 Chr.5: 55256548-55256548 on Build GRCh38
rs1034993017 Chr.5: 55256511-55256511 on Build GRCh38
rs550237034 Chr.5: 55256553-55256553 on Build GRCh38
rs377731810 Chr.5: 55256560-55256560 on Build GRCh38

View Full Product Details