Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58712553-58712818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYAT
Entrez Gene ID
10249
Gene Name
glycine-N-acyltransferase
Gene Aliases
ACGNAT, CAT, GAT
UniGene
Hs.145384
Gene Chromosome Location
Chr.11: 58708757-58731974 on Build GRCh38
Transcripts
NM_005838.3 NM_201648.2 XM_017017087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416017 c.301C>A p.H101N Chr.11: 58712775-58712775 on Build GRCh38
231247 c.272G>A p.G91E Chr.11: 58712804-58712804 on Build GRCh38
4034212 c.269T>C p.L90P Chr.11: 58712807-58712807 on Build GRCh38
3450048 c.288C>T p.I96I Chr.11: 58712788-58712788 on Build GRCh38
6133523 c.306A>G p.L102L Chr.11: 58712770-58712770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868606378 Chr.11: 58712748-58712748 on Build GRCh38
rs768971264 Chr.11: 58712761-58712761 on Build GRCh38
rs748514292 Chr.11: 58712775-58712775 on Build GRCh38
rs969574326 Chr.11: 58712580-58712580 on Build GRCh38
rs76187651 Chr.11: 58712745-58712745 on Build GRCh38
rs139286707 Chr.11: 58712604-58712604 on Build GRCh38
rs992467185 Chr.11: 58712629-58712629 on Build GRCh38
rs2509908 Chr.11: 58712682-58712682 on Build GRCh38
rs548028778 Chr.11: 58712652-58712652 on Build GRCh38
rs1010338698 Chr.11: 58712616-58712616 on Build GRCh38
rs778571237 Chr.11: 58712583-58712583 on Build GRCh38
rs905569165 Chr.11: 58712586-58712586 on Build GRCh38
rs1019887188 Chr.11: 58712635-58712635 on Build GRCh38
rs959727725 Chr.11: 58712614-58712614 on Build GRCh38
rs376149234 Chr.11: 58712712-58712712 on Build GRCh38
rs555283386 Chr.11: 58712722-58712722 on Build GRCh38
rs777004801 Chr.11: 58712765-58712765 on Build GRCh38
rs539199116 Chr.11: 58712624-58712624 on Build GRCh38
rs755812519 Chr.11: 58712734-58712734 on Build GRCh38
rs763072250 Chr.11: 58712793-58712793 on Build GRCh38
rs537480013 Chr.11: 58712776-58712776 on Build GRCh38
rs566327824 Chr.11: 58712665-58712665 on Build GRCh38
rs978985806 Chr.11: 58712669-58712669 on Build GRCh38
rs751695296 Chr.11: 58712632-58712632 on Build GRCh38
rs773369748 Chr.11: 58712784-58712784 on Build GRCh38
rs372984833 Chr.11: 58712741-58712741 on Build GRCh38
rs967537072 Chr.11: 58712651-58712651 on Build GRCh38
rs781137498 Chr.11: 58712716-58712716 on Build GRCh38
rs777340171 Chr.11: 58712739-58712739 on Build GRCh38
rs747520674 Chr.11: 58712749-58712749 on Build GRCh38
rs983252473 Chr.11: 58712772-58712772 on Build GRCh38
rs986073010 Chr.11: 58712701-58712701 on Build GRCh38
rs748826338 Chr.11: 58712740-58712740 on Build GRCh38
rs766079626 Chr.11: 58712724-58712724 on Build GRCh38
rs751602266 Chr.11: 58712709-58712709 on Build GRCh38
rs115534061 Chr.11: 58712605-58712605 on Build GRCh38

View Full Product Details