Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70429642-70429883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD2
Entrez Gene ID
54857
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Gene Aliases
GDE3, OBDPF
UniGene
Hs.438712
Gene Chromosome Location
Chr.X: 70423031-70433391 on Build GRCh38
Transcripts
NM_001171191.1 NM_001171192.1 NM_001171193.1 NM_017711.3 XM_011530977.1 XM_017029614.1 XM_011531081.2 XM_011531082.2

Gene Symbol
LOC105373244
Entrez Gene ID
105373244
Gene Name
uncharacterized LOC105373244
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.X: 70427358-70436800 on Build GRCh38
Transcripts
NM_001171191.1 NM_001171192.1 NM_001171193.1 NM_017711.3 XM_011530977.1 XM_017029614.1 XM_011531081.2 XM_011531082.2

SNP Detail

SNP ID Chromosome Location
rs766267643 Chr.X: 70429681-70429681 on Build GRCh38
rs778000608 Chr.X: 70429784-70429784 on Build GRCh38
rs757087205 Chr.X: 70429693-70429693 on Build GRCh38
rs142847810 Chr.X: 70429819-70429819 on Build GRCh38
rs774966799 Chr.X: 70429807-70429807 on Build GRCh38
rs749604543 Chr.X: 70429794-70429794 on Build GRCh38
rs184886139 Chr.X: 70429802-70429802 on Build GRCh38
rs751318015 Chr.X: 70429683-70429683 on Build GRCh38
rs929668981 Chr.X: 70429791-70429791 on Build GRCh38
rs779316426 Chr.X: 70429800-70429800 on Build GRCh38
rs915160891 Chr.X: 70429723-70429723 on Build GRCh38
rs776115679 Chr.X: 70429842-70429845 on Build GRCh38
rs774042784 Chr.X: 70429849-70429849 on Build GRCh38
rs755208606 Chr.X: 70429798-70429798 on Build GRCh38
rs749405886 Chr.X: 70429795-70429795 on Build GRCh38
rs747408478 Chr.X: 70429817-70429817 on Build GRCh38
rs745847245 Chr.X: 70429852-70429852 on Build GRCh38
rs1012683483 Chr.X: 70429856-70429856 on Build GRCh38
rs1056780524 Chr.X: 70429679-70429679 on Build GRCh38
rs763707430 Chr.X: 70429841-70429841 on Build GRCh38
rs767423160 Chr.X: 70429700-70429700 on Build GRCh38
rs750390154 Chr.X: 70429730-70429730 on Build GRCh38
rs1003373180 Chr.X: 70429804-70429804 on Build GRCh38
rs748500485 Chr.X: 70429801-70429801 on Build GRCh38
rs763760397 Chr.X: 70429676-70429676 on Build GRCh38
rs753676131 Chr.X: 70429743-70429743 on Build GRCh38
rs768216482 Chr.X: 70429822-70429822 on Build GRCh38
rs774814271 Chr.X: 70429820-70429820 on Build GRCh38

View Full Product Details