Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179944953-179945226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC22
Entrez Gene ID
57703
Gene Name
CWC22 homolog, spliceosome-associated protein
Gene Aliases
EIF4GL, NCM, fSAPb
UniGene
Hs.311363
Gene Chromosome Location
Chr.2: 179944876-180007359 on Build GRCh38
Transcripts
NM_020943.2 XM_005246726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2710228 c.2692C>T p.R898W Chr.2: 179945164-179945164 on Build GRCh38
1482524 c.2666C>T p.S889F Chr.2: 179945190-179945190 on Build GRCh38
3425825 c.2695C>T p.R899* Chr.2: 179945161-179945161 on Build GRCh38
5658821 c.2638G>C p.E880Q Chr.2: 179945218-179945218 on Build GRCh38
280233 c.2634A>G p.R878R Chr.2: 179945222-179945222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776538518 Chr.2: 179945117-179945117 on Build GRCh38
rs926527849 Chr.2: 179945114-179945114 on Build GRCh38
rs946046045 Chr.2: 179945039-179945039 on Build GRCh38
rs751068671 Chr.2: 179945163-179945163 on Build GRCh38
rs931170379 Chr.2: 179945119-179945119 on Build GRCh38
rs769782797 Chr.2: 179945132-179945132 on Build GRCh38
rs754181079 Chr.2: 179945029-179945029 on Build GRCh38
rs373464895 Chr.2: 179945079-179945079 on Build GRCh38
rs750870010 Chr.2: 179945169-179945169 on Build GRCh38
rs907427878 Chr.2: 179945049-179945049 on Build GRCh38
rs761999005 Chr.2: 179945126-179945126 on Build GRCh38
rs746406630 Chr.2: 179945082-179945082 on Build GRCh38
rs763700302 Chr.2: 179945095-179945095 on Build GRCh38
rs766266576 Chr.2: 179945142-179945142 on Build GRCh38
rs1001017750 Chr.2: 179945061-179945061 on Build GRCh38
rs768932564 Chr.2: 179945112-179945112 on Build GRCh38
rs762999399 Chr.2: 179945141-179945141 on Build GRCh38
rs773159239 Chr.2: 179945135-179945135 on Build GRCh38
rs1054548857 Chr.2: 179944978-179944979 on Build GRCh38
rs996496456 Chr.2: 179945179-179945179 on Build GRCh38
rs988208423 Chr.2: 179945077-179945077 on Build GRCh38
rs1056358626 Chr.2: 179945095-179945096 on Build GRCh38
rs752315626 Chr.2: 179945161-179945161 on Build GRCh38
rs573098807 Chr.2: 179945160-179945160 on Build GRCh38
rs756821579 Chr.2: 179945172-179945172 on Build GRCh38
rs779428205 Chr.2: 179945080-179945080 on Build GRCh38
rs751470890 Chr.2: 179945146-179945146 on Build GRCh38
rs1032469114 Chr.2: 179945062-179945062 on Build GRCh38
rs932026071 Chr.2: 179944976-179944976 on Build GRCh38
rs1029224257 Chr.2: 179944989-179944989 on Build GRCh38
rs953537787 Chr.2: 179945041-179945041 on Build GRCh38
rs777154447 Chr.2: 179945164-179945164 on Build GRCh38
rs759237563 Chr.2: 179945154-179945154 on Build GRCh38
rs957000949 Chr.2: 179945075-179945075 on Build GRCh38
rs185736881 Chr.2: 179944978-179944978 on Build GRCh38
rs780407677 Chr.2: 179945096-179945096 on Build GRCh38
rs377091525 Chr.2: 179945099-179945099 on Build GRCh38
rs762613746 Chr.2: 179945071-179945071 on Build GRCh38

View Full Product Details