Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58946997-58947224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL1
Entrez Gene ID
92292
Gene Name
glycine-N-acyltransferase like 1
Gene Aliases
GATF-C, GNAT
UniGene
Hs.38085
Gene Chromosome Location
Chr.11: 58927629-58957074 on Build GRCh38
Transcripts
NM_001220494.2 NM_001220496.2 NM_080661.4 XM_011545354.2 XM_011545355.2 XM_011545356.2 XM_011545357.2 XM_011545358.2 XM_011545359.2 NR_033853.2

Gene Symbol
LOC283194
Entrez Gene ID
283194
Gene Name
uncharacterized LOC283194
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 58933643-59058452 on Build GRCh38
Transcripts
NM_001220494.2 NM_001220496.2 NM_080661.4 XM_011545354.2 XM_011545355.2 XM_011545356.2 XM_011545357.2 XM_011545358.2 XM_011545359.2 NR_033853.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429319 c.3G>A p.M1I Chr.11: 58947090-58947090 on Build GRCh38
928926 c.30G>A p.L10L Chr.11: 58947117-58947117 on Build GRCh38
4905241 c.149-1G>A p.? Chr.11: 58947045-58947045 on Build GRCh38
6837921 c.35C>A p.A12D Chr.11: 58947122-58947122 on Build GRCh38
348472 c.33G>T p.L11L Chr.11: 58947120-58947120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764585487 Chr.11: 58947176-58947176 on Build GRCh38
rs201056557 Chr.11: 58947030-58947030 on Build GRCh38
rs367748078 Chr.11: 58947103-58947103 on Build GRCh38
rs563291054 Chr.11: 58947195-58947195 on Build GRCh38
rs772697858 Chr.11: 58947127-58947127 on Build GRCh38
rs375279836 Chr.11: 58947116-58947116 on Build GRCh38
rs192751674 Chr.11: 58947101-58947101 on Build GRCh38
rs780665861 Chr.11: 58947104-58947104 on Build GRCh38
rs754096489 Chr.11: 58947065-58947065 on Build GRCh38
rs112396597 Chr.11: 58947037-58947037 on Build GRCh38
rs957766431 Chr.11: 58947044-58947044 on Build GRCh38
rs2154968 Chr.11: 58947160-58947160 on Build GRCh38
rs200383788 Chr.11: 58947032-58947032 on Build GRCh38
rs750508110 Chr.11: 58947090-58947090 on Build GRCh38
rs758481995 Chr.11: 58947196-58947196 on Build GRCh38
rs776154089 Chr.11: 58947133-58947133 on Build GRCh38
rs765443710 Chr.11: 58947193-58947193 on Build GRCh38
rs371493000 Chr.11: 58947108-58947108 on Build GRCh38
rs772751412 Chr.11: 58947122-58947122 on Build GRCh38
rs764354434 Chr.11: 58947057-58947057 on Build GRCh38
rs762062535 Chr.11: 58947192-58947192 on Build GRCh38
rs779025892 Chr.11: 58947086-58947086 on Build GRCh38
rs762622949 Chr.11: 58947126-58947126 on Build GRCh38
rs188134197 Chr.11: 58947033-58947033 on Build GRCh38
rs780024562 Chr.11: 58947097-58947097 on Build GRCh38
rs769383337 Chr.11: 58947121-58947121 on Build GRCh38
rs542320563 Chr.11: 58947178-58947178 on Build GRCh38
rs989688049 Chr.11: 58947061-58947061 on Build GRCh38
rs761055707 Chr.11: 58947157-58947157 on Build GRCh38
rs758326202 Chr.11: 58947091-58947091 on Build GRCh38
rs757246287 Chr.11: 58947079-58947079 on Build GRCh38
rs1003262193 Chr.11: 58947139-58947139 on Build GRCh38
rs753110546 Chr.11: 58947035-58947035 on Build GRCh38
rs1035707695 Chr.11: 58947171-58947171 on Build GRCh38

View Full Product Details