Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10971022-10971283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
UniGene
Hs.35490
Gene Chromosome Location
Chr.16: 10944488-11193272 on Build GRCh38
Transcripts
NM_001243403.1 NM_015226.2 XM_005255210.1 XM_005255211.1 XM_005255213.1 XM_005255214.1 XM_005255215.3 XM_005255216.1 XM_006720870.3 XM_011522434.1 XM_011522435.1 XM_011522436.2 XM_011522437.2 XM_011522438.2 XM_011522439.2 XM_011522440.2 XM_017023089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630114 c.523G>A p.A175T Chr.16: 10971155-10971155 on Build GRCh38
6143553 c.597A>T p.K199N Chr.16: 10971229-10971229 on Build GRCh38
4640034 c.513G>A p.L171L Chr.16: 10971145-10971145 on Build GRCh38
4635933 c.552C>T p.S184S Chr.16: 10971184-10971184 on Build GRCh38
7232359 c.537C>G p.F179L Chr.16: 10971169-10971169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201695473 Chr.16: 10971244-10971244 on Build GRCh38
rs568057166 Chr.16: 10971249-10971249 on Build GRCh38
rs964084227 Chr.16: 10971163-10971163 on Build GRCh38
rs199890590 Chr.16: 10971258-10971258 on Build GRCh38
rs759604074 Chr.16: 10971172-10971172 on Build GRCh38
rs201211737 Chr.16: 10971259-10971259 on Build GRCh38
rs199868443 Chr.16: 10971106-10971106 on Build GRCh38
rs774299916 Chr.16: 10971171-10971171 on Build GRCh38
rs763222619 Chr.16: 10971231-10971232 on Build GRCh38
rs567150021 Chr.16: 10971257-10971257 on Build GRCh38
rs369393972 Chr.16: 10971086-10971086 on Build GRCh38
rs867691682 Chr.16: 10971207-10971207 on Build GRCh38
rs747798193 Chr.16: 10971117-10971117 on Build GRCh38
rs753032767 Chr.16: 10971200-10971200 on Build GRCh38
rs763394459 Chr.16: 10971140-10971140 on Build GRCh38
rs192972852 Chr.16: 10971233-10971233 on Build GRCh38
rs72650684 Chr.16: 10971141-10971141 on Build GRCh38
rs200366711 Chr.16: 10971093-10971093 on Build GRCh38
rs1018285498 Chr.16: 10971139-10971139 on Build GRCh38
rs757175950 Chr.16: 10971076-10971076 on Build GRCh38
rs776327190 Chr.16: 10971109-10971109 on Build GRCh38
rs779970758 Chr.16: 10971236-10971236 on Build GRCh38
rs773546785 Chr.16: 10971136-10971136 on Build GRCh38
rs778844500 Chr.16: 10971082-10971082 on Build GRCh38
rs758331636 Chr.16: 10971087-10971087 on Build GRCh38
rs188575867 Chr.16: 10971059-10971059 on Build GRCh38
rs758294558 Chr.16: 10971234-10971234 on Build GRCh38
rs771431541 Chr.16: 10971151-10971151 on Build GRCh38
rs530438782 Chr.16: 10971215-10971215 on Build GRCh38
rs200404847 Chr.16: 10971094-10971094 on Build GRCh38
rs909902243 Chr.16: 10971143-10971143 on Build GRCh38
rs770186127 Chr.16: 10971123-10971123 on Build GRCh38
rs760947013 Chr.16: 10971211-10971211 on Build GRCh38
rs200906965 Chr.16: 10971243-10971243 on Build GRCh38
rs371603009 Chr.16: 10971261-10971261 on Build GRCh38
rs767691187 Chr.16: 10971176-10971176 on Build GRCh38
rs201388822 Chr.16: 10971105-10971105 on Build GRCh38

View Full Product Details