Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1313022-1313263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424020 c.1110C>A p.L370L Chr.1: 1313056-1313056 on Build GRCh38
6197350 c.1141C>T p.R381W Chr.1: 1313043-1313043 on Build GRCh38
3984146 c.1049T>C p.M350T Chr.1: 1313117-1313117 on Build GRCh38
5424021 c.1128C>A p.L376L Chr.1: 1313056-1313056 on Build GRCh38
6634873 c.1142G>A p.R381Q Chr.1: 1313042-1313042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541205411 Chr.1: 1313241-1313241 on Build GRCh38
rs778939217 Chr.1: 1313131-1313131 on Build GRCh38
rs376193505 Chr.1: 1313134-1313134 on Build GRCh38
rs750944542 Chr.1: 1313113-1313113 on Build GRCh38
rs768724959 Chr.1: 1313063-1313063 on Build GRCh38
rs898732953 Chr.1: 1313188-1313188 on Build GRCh38
rs1029646516 Chr.1: 1313203-1313203 on Build GRCh38
rs74045438 Chr.1: 1313206-1313206 on Build GRCh38
rs749570256 Chr.1: 1313127-1313127 on Build GRCh38
rs997283135 Chr.1: 1313199-1313199 on Build GRCh38
rs752173663 Chr.1: 1313236-1313239 on Build GRCh38
rs765080194 Chr.1: 1313187-1313187 on Build GRCh38
rs527463409 Chr.1: 1313240-1313240 on Build GRCh38
rs773862725 Chr.1: 1313169-1313169 on Build GRCh38
rs764982365 Chr.1: 1313088-1313088 on Build GRCh38
rs772115268 Chr.1: 1313135-1313135 on Build GRCh38
rs751228573 Chr.1: 1313092-1313092 on Build GRCh38
rs775418738 Chr.1: 1313153-1313153 on Build GRCh38
rs776859024 Chr.1: 1313146-1313146 on Build GRCh38
rs140151309 Chr.1: 1313104-1313104 on Build GRCh38
rs563141128 Chr.1: 1313089-1313089 on Build GRCh38
rs529799687 Chr.1: 1313181-1313181 on Build GRCh38
rs768561022 Chr.1: 1313158-1313158 on Build GRCh38
rs771049158 Chr.1: 1313091-1313091 on Build GRCh38
rs780616170 Chr.1: 1313056-1313056 on Build GRCh38
rs755386598 Chr.1: 1313128-1313128 on Build GRCh38
rs1050321286 Chr.1: 1313197-1313197 on Build GRCh38
rs998906460 Chr.1: 1313207-1313207 on Build GRCh38
rs747119335 Chr.1: 1313149-1313149 on Build GRCh38
rs769669296 Chr.1: 1313150-1313150 on Build GRCh38
rs9725147 Chr.1: 1313219-1313219 on Build GRCh38
rs772257638 Chr.1: 1313077-1313077 on Build GRCh38
rs762829023 Chr.1: 1313090-1313090 on Build GRCh38
rs780741157 Chr.1: 1313116-1313116 on Build GRCh38
rs990328110 Chr.1: 1313234-1313234 on Build GRCh38
rs142980334 Chr.1: 1313087-1313087 on Build GRCh38
rs917696621 Chr.1: 1313132-1313132 on Build GRCh38
rs746446373 Chr.1: 1313043-1313043 on Build GRCh38
rs762585928 Chr.1: 1313155-1313155 on Build GRCh38
rs756534506 Chr.1: 1313050-1313050 on Build GRCh38
rs1031845888 Chr.1: 1313217-1313217 on Build GRCh38
rs776534078 Chr.1: 1313085-1313085 on Build GRCh38
rs762958897 Chr.1: 1313242-1313242 on Build GRCh38
rs756719002 Chr.1: 1313103-1313103 on Build GRCh38
rs548248819 Chr.1: 1313198-1313198 on Build GRCh38
rs774658242 Chr.1: 1313065-1313065 on Build GRCh38
rs748121981 Chr.1: 1313071-1313071 on Build GRCh38
rs899740247 Chr.1: 1313193-1313193 on Build GRCh38
rs761576095 Chr.1: 1313171-1313171 on Build GRCh38

View Full Product Details