Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75003386-75003647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477773 c.331A>T p.K111* Chr.14: 75003597-75003597 on Build GRCh38
3793864 c.293G>A p.G98E Chr.14: 75003559-75003559 on Build GRCh38
433375 c.301G>A p.D101N Chr.14: 75003567-75003567 on Build GRCh38
5824579 c.367A>T p.S123C Chr.14: 75003633-75003633 on Build GRCh38
5848544 c.344C>T p.S115F Chr.14: 75003610-75003610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777252700 Chr.14: 75003500-75003500 on Build GRCh38
rs780391000 Chr.14: 75003434-75003434 on Build GRCh38
rs762604029 Chr.14: 75003543-75003543 on Build GRCh38
rs770508411 Chr.14: 75003518-75003518 on Build GRCh38
rs368133564 Chr.14: 75003564-75003564 on Build GRCh38
rs756670980 Chr.14: 75003429-75003429 on Build GRCh38
rs967630584 Chr.14: 75003491-75003491 on Build GRCh38
rs370483997 Chr.14: 75003591-75003591 on Build GRCh38
rs767584462 Chr.14: 75003418-75003418 on Build GRCh38
rs755393155 Chr.14: 75003440-75003440 on Build GRCh38
rs759621968 Chr.14: 75003599-75003599 on Build GRCh38
rs746678040 Chr.14: 75003512-75003512 on Build GRCh38
rs918318919 Chr.14: 75003464-75003464 on Build GRCh38
rs17849324 Chr.14: 75003565-75003565 on Build GRCh38
rs775335483 Chr.14: 75003537-75003537 on Build GRCh38
rs997727777 Chr.14: 75003501-75003501 on Build GRCh38
rs1030584724 Chr.14: 75003506-75003506 on Build GRCh38
rs374195020 Chr.14: 75003620-75003620 on Build GRCh38
rs776342453 Chr.14: 75003522-75003522 on Build GRCh38
rs183116394 Chr.14: 75003437-75003437 on Build GRCh38
rs769894290 Chr.14: 75003532-75003532 on Build GRCh38
rs368343040 Chr.14: 75003621-75003621 on Build GRCh38
rs373906805 Chr.14: 75003550-75003550 on Build GRCh38
rs1020267453 Chr.14: 75003468-75003468 on Build GRCh38
rs768864027 Chr.14: 75003521-75003522 on Build GRCh38
rs750868104 Chr.14: 75003424-75003424 on Build GRCh38
rs779349624 Chr.14: 75003611-75003611 on Build GRCh38
rs768524274 Chr.14: 75003544-75003544 on Build GRCh38
rs751826162 Chr.14: 75003407-75003407 on Build GRCh38
rs766843755 Chr.14: 75003577-75003577 on Build GRCh38
rs751159755 Chr.14: 75003612-75003612 on Build GRCh38
rs750568077 Chr.14: 75003557-75003557 on Build GRCh38
rs200073232 Chr.14: 75003411-75003411 on Build GRCh38
rs762068601 Chr.14: 75003547-75003547 on Build GRCh38
rs755232669 Chr.14: 75003608-75003608 on Build GRCh38
rs373932168 Chr.14: 75003528-75003528 on Build GRCh38
rs376049144 Chr.14: 75003549-75003549 on Build GRCh38

View Full Product Details