Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25483392-25483617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
UniGene
Hs.33366
Gene Chromosome Location
Chr.8: 25458569-25511308 on Build GRCh38
Transcripts
NM_001317906.1 NM_001317907.1 NM_152562.3 XM_006716294.2 XM_011544415.1 XM_011544416.2 XM_011544417.2 XM_011544418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3374869 c.1053T>G p.Y351* Chr.8: 25483419-25483419 on Build GRCh38
4636785 c.1050C>G p.L350L Chr.8: 25483416-25483416 on Build GRCh38
3647611 c.1059T>C p.D353D Chr.8: 25483425-25483425 on Build GRCh38
3647612 c.1073C>T p.P358L Chr.8: 25483439-25483439 on Build GRCh38
5467401 c.1074G>A p.P358P Chr.8: 25483440-25483440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017640286 Chr.8: 25483568-25483568 on Build GRCh38
rs529716489 Chr.8: 25483581-25483581 on Build GRCh38
rs144835235 Chr.8: 25483440-25483440 on Build GRCh38
rs761131272 Chr.8: 25483459-25483459 on Build GRCh38
rs767430932 Chr.8: 25483530-25483530 on Build GRCh38
rs776051762 Chr.8: 25483452-25483452 on Build GRCh38
rs974521049 Chr.8: 25483435-25483435 on Build GRCh38
rs1009312496 Chr.8: 25483517-25483517 on Build GRCh38
rs796620653 Chr.8: 25483572-25483572 on Build GRCh38
rs141028295 Chr.8: 25483422-25483422 on Build GRCh38
rs934628044 Chr.8: 25483564-25483564 on Build GRCh38
rs759537889 Chr.8: 25483526-25483526 on Build GRCh38
rs772699869 Chr.8: 25483446-25483446 on Build GRCh38
rs762463563 Chr.8: 25483473-25483473 on Build GRCh38
rs964264386 Chr.8: 25483577-25483577 on Build GRCh38
rs148572164 Chr.8: 25483471-25483471 on Build GRCh38
rs771078018 Chr.8: 25483428-25483428 on Build GRCh38
rs772948782 Chr.8: 25483467-25483467 on Build GRCh38
rs965768961 Chr.8: 25483434-25483434 on Build GRCh38
rs367833985 Chr.8: 25483461-25483461 on Build GRCh38
rs988790944 Chr.8: 25483569-25483569 on Build GRCh38
rs144887977 Chr.8: 25483439-25483439 on Build GRCh38
rs765961468 Chr.8: 25483490-25483490 on Build GRCh38
rs562374338 Chr.8: 25483483-25483483 on Build GRCh38
rs751000005 Chr.8: 25483516-25483516 on Build GRCh38

View Full Product Details