Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114245881-114246154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
UniGene
Hs.374127
Gene Chromosome Location
Chr.13: 114234845-114272675 on Build GRCh38
Transcripts
NM_001078645.1 NM_001318517.1 NM_001318518.1 NM_003903.3 XM_005266206.1 XM_006719992.1 XM_006719993.1 XM_011534851.1 XM_011534853.1 XM_017020830.1 XM_017020831.1 XM_017020832.1 XM_017020833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4839331 c.848-1G>T p.? Chr.13: 114245999-114245999 on Build GRCh38
1322957 c.859C>G p.L287V Chr.13: 114246011-114246011 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757531635 Chr.13: 114246069-114246069 on Build GRCh38
rs1013506246 Chr.13: 114246117-114246117 on Build GRCh38
rs924624729 Chr.13: 114245914-114245914 on Build GRCh38
rs745940569 Chr.13: 114246088-114246088 on Build GRCh38
rs1031179385 Chr.13: 114246116-114246116 on Build GRCh38
rs147824768 Chr.13: 114245964-114245964 on Build GRCh38
rs1007136033 Chr.13: 114246058-114246058 on Build GRCh38
rs201167565 Chr.13: 114246088-114246089 on Build GRCh38
rs762541081 Chr.13: 114246006-114246006 on Build GRCh38
rs773728556 Chr.13: 114246076-114246077 on Build GRCh38
rs758541666 Chr.13: 114246049-114246049 on Build GRCh38
rs772851654 Chr.13: 114245991-114245991 on Build GRCh38
rs777025165 Chr.13: 114246075-114246075 on Build GRCh38
rs17291229 Chr.13: 114245941-114245941 on Build GRCh38
rs888798765 Chr.13: 114246052-114246052 on Build GRCh38
rs751044276 Chr.13: 114246011-114246011 on Build GRCh38
rs371610353 Chr.13: 114246099-114246099 on Build GRCh38
rs770101903 Chr.13: 114246075-114246076 on Build GRCh38
rs994596654 Chr.13: 114245992-114245992 on Build GRCh38
rs372936394 Chr.13: 114245957-114245957 on Build GRCh38
rs370057318 Chr.13: 114246076-114246076 on Build GRCh38
rs897602997 Chr.13: 114245976-114245976 on Build GRCh38
rs749479640 Chr.13: 114246063-114246063 on Build GRCh38
rs535891590 Chr.13: 114245982-114245982 on Build GRCh38
rs779320626 Chr.13: 114246078-114246078 on Build GRCh38
rs767436979 Chr.13: 114246037-114246037 on Build GRCh38
rs17337996 Chr.13: 114246059-114246059 on Build GRCh38
rs529415431 Chr.13: 114246112-114246112 on Build GRCh38
rs370534631 Chr.13: 114245970-114245970 on Build GRCh38
rs139331334 Chr.13: 114246018-114246018 on Build GRCh38
rs768181031 Chr.13: 114246066-114246071 on Build GRCh38
rs931828426 Chr.13: 114245942-114245942 on Build GRCh38
rs765824773 Chr.13: 114246007-114246007 on Build GRCh38
rs74116851 Chr.13: 114246075-114246075 on Build GRCh38
rs17291222 Chr.13: 114245931-114245931 on Build GRCh38
rs754521696 Chr.13: 114246032-114246032 on Build GRCh38
rs958912546 Chr.13: 114246114-114246114 on Build GRCh38
rs777460721 Chr.13: 114246060-114246060 on Build GRCh38
rs947951558 Chr.13: 114245934-114245934 on Build GRCh38
rs776207753 Chr.13: 114245965-114245965 on Build GRCh38
rs746741458 Chr.13: 114246006-114246009 on Build GRCh38
rs369349089 Chr.13: 114245956-114245956 on Build GRCh38
rs989408482 Chr.13: 114245918-114245918 on Build GRCh38
rs532283869 Chr.13: 114245953-114245953 on Build GRCh38

View Full Product Details