Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26883416-26883665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA5
Entrez Gene ID
2558
Gene Name
gamma-aminobutyric acid type A receptor alpha5 subunit
Gene Aliases
-
UniGene
Hs.24969
Gene Chromosome Location
Chr.15: 26866719-26949210 on Build GRCh38
Transcripts
NM_000810.3 NM_001165037.1 XM_005268258.2 XM_006720459.2 XM_017022055.1 XM_017022056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1734041 c.361A>T p.N121Y Chr.15: 26883421-26883421 on Build GRCh38
1242302 c.411C>T p.N137N Chr.15: 26883471-26883471 on Build GRCh38
700321 c.427G>C p.A143P Chr.15: 26883487-26883487 on Build GRCh38
183776 c.377G>A p.S126N Chr.15: 26883437-26883437 on Build GRCh38
6046067 c.497+6C>T p.? Chr.15: 26883563-26883563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996421944 Chr.15: 26883640-26883640 on Build GRCh38
rs947692047 Chr.15: 26883614-26883614 on Build GRCh38
rs768455924 Chr.15: 26883456-26883456 on Build GRCh38
rs747836136 Chr.15: 26883586-26883586 on Build GRCh38
rs778645917 Chr.15: 26883459-26883459 on Build GRCh38
rs371590711 Chr.15: 26883534-26883534 on Build GRCh38
rs373457164 Chr.15: 26883523-26883523 on Build GRCh38
rs777312866 Chr.15: 26883602-26883602 on Build GRCh38
rs750378125 Chr.15: 26883554-26883554 on Build GRCh38
rs780101118 Chr.15: 26883575-26883575 on Build GRCh38
rs760055096 Chr.15: 26883512-26883512 on Build GRCh38
rs201231068 Chr.15: 26883484-26883484 on Build GRCh38
rs113499558 Chr.15: 26883582-26883582 on Build GRCh38
rs778489846 Chr.15: 26883583-26883583 on Build GRCh38
rs764304732 Chr.15: 26883531-26883531 on Build GRCh38
rs992811669 Chr.15: 26883609-26883609 on Build GRCh38
rs200583728 Chr.15: 26883486-26883486 on Build GRCh38
rs978194844 Chr.15: 26883587-26883587 on Build GRCh38
rs984556074 Chr.15: 26883629-26883629 on Build GRCh38
rs770188153 Chr.15: 26883517-26883517 on Build GRCh38
rs961024879 Chr.15: 26883605-26883605 on Build GRCh38
rs749042436 Chr.15: 26883436-26883436 on Build GRCh38
rs902073158 Chr.15: 26883626-26883626 on Build GRCh38
rs908934936 Chr.15: 26883637-26883637 on Build GRCh38
rs761854701 Chr.15: 26883538-26883538 on Build GRCh38
rs777074772 Chr.15: 26883510-26883510 on Build GRCh38
rs889779772 Chr.15: 26883506-26883506 on Build GRCh38
rs776191953 Chr.15: 26883583-26883609 on Build GRCh38
rs539255264 Chr.15: 26883581-26883581 on Build GRCh38
rs767604007 Chr.15: 26883540-26883540 on Build GRCh38
rs866557200 Chr.15: 26883520-26883520 on Build GRCh38
rs1029702598 Chr.15: 26883491-26883491 on Build GRCh38
rs968198759 Chr.15: 26883535-26883535 on Build GRCh38
rs746686415 Chr.15: 26883571-26883571 on Build GRCh38
rs372061541 Chr.15: 26883462-26883462 on Build GRCh38
rs374917841 Chr.15: 26883526-26883526 on Build GRCh38
rs940514107 Chr.15: 26883641-26883641 on Build GRCh38
rs756143877 Chr.15: 26883567-26883567 on Build GRCh38
rs771519220 Chr.15: 26883598-26883598 on Build GRCh38
rs573388684 Chr.15: 26883610-26883610 on Build GRCh38
rs753693567 Chr.15: 26883579-26883579 on Build GRCh38
rs1031214425 Chr.15: 26883594-26883594 on Build GRCh38
rs779783248 Chr.15: 26883471-26883474 on Build GRCh38

View Full Product Details