Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37783855-37784128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC3
Entrez Gene ID
51010
Gene Name
exosome component 3
Gene Aliases
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10
UniGene
Hs.602571
Gene Chromosome Location
Chr.9: 37779714-37785092 on Build GRCh38
Transcripts
NM_001002269.2 NM_016042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334532 c.389A>G p.K130R Chr.9: 37783999-37783999 on Build GRCh38
4912147 c.414A>G p.P138P Chr.9: 37783974-37783974 on Build GRCh38
4682067 c.473A>G p.Q158R Chr.9: 37783915-37783915 on Build GRCh38
3375247 c.387C>G p.F129L Chr.9: 37784001-37784001 on Build GRCh38
50583 c.372A>T p.K124N Chr.9: 37784016-37784016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773161484 Chr.9: 37784010-37784010 on Build GRCh38
rs749142446 Chr.9: 37784058-37784059 on Build GRCh38
rs919744989 Chr.9: 37783905-37783905 on Build GRCh38
rs999232158 Chr.9: 37784041-37784041 on Build GRCh38
rs760424055 Chr.9: 37783883-37783883 on Build GRCh38
rs1044124443 Chr.9: 37783996-37783996 on Build GRCh38
rs768414644 Chr.9: 37783890-37783890 on Build GRCh38
rs776611730 Chr.9: 37783895-37783895 on Build GRCh38
rs768507877 Chr.9: 37783994-37783994 on Build GRCh38
rs146985065 Chr.9: 37784077-37784083 on Build GRCh38
rs573975449 Chr.9: 37783967-37783967 on Build GRCh38
rs753133011 Chr.9: 37784073-37784073 on Build GRCh38
rs751776878 Chr.9: 37783955-37783955 on Build GRCh38
rs138169215 Chr.9: 37784060-37784060 on Build GRCh38
rs766461372 Chr.9: 37784054-37784054 on Build GRCh38
rs746439813 Chr.9: 37784088-37784088 on Build GRCh38
rs145622193 Chr.9: 37784027-37784027 on Build GRCh38
rs762680724 Chr.9: 37784045-37784047 on Build GRCh38
rs761749070 Chr.9: 37784082-37784082 on Build GRCh38
rs768439296 Chr.9: 37784073-37784082 on Build GRCh38
rs373193552 Chr.9: 37784000-37784000 on Build GRCh38
rs774069700 Chr.9: 37784078-37784083 on Build GRCh38
rs140345562 Chr.9: 37783920-37783920 on Build GRCh38
rs764545296 Chr.9: 37784082-37784082 on Build GRCh38
rs756503706 Chr.9: 37784079-37784079 on Build GRCh38
rs1002846587 Chr.9: 37783992-37783992 on Build GRCh38
rs145464176 Chr.9: 37783940-37783940 on Build GRCh38
rs780777501 Chr.9: 37784093-37784093 on Build GRCh38
rs779619010 Chr.9: 37783987-37783987 on Build GRCh38
rs141138948 Chr.9: 37783993-37783993 on Build GRCh38
rs138085418 Chr.9: 37783958-37783958 on Build GRCh38
rs747883936 Chr.9: 37784001-37784001 on Build GRCh38
rs1044852285 Chr.9: 37783947-37783947 on Build GRCh38
rs778372165 Chr.9: 37783969-37783969 on Build GRCh38
rs387907195 Chr.9: 37783973-37783973 on Build GRCh38
rs776602138 Chr.9: 37784089-37784090 on Build GRCh38
rs375827861 Chr.9: 37784069-37784069 on Build GRCh38
rs756702077 Chr.9: 37783966-37783966 on Build GRCh38
rs181508905 Chr.9: 37783899-37783899 on Build GRCh38
rs750485929 Chr.9: 37783917-37783917 on Build GRCh38
rs903357748 Chr.9: 37784005-37784005 on Build GRCh38
rs778400947 Chr.9: 37783906-37783906 on Build GRCh38
rs202059195 Chr.9: 37784083-37784083 on Build GRCh38
rs781685540 Chr.9: 37783959-37783959 on Build GRCh38
rs200453185 Chr.9: 37784099-37784099 on Build GRCh38
rs769746459 Chr.9: 37784009-37784009 on Build GRCh38
rs751936620 Chr.9: 37783997-37783997 on Build GRCh38
rs911113863 Chr.9: 37783888-37783888 on Build GRCh38
rs767436365 Chr.9: 37784082-37784083 on Build GRCh38
rs779614399 Chr.9: 37784087-37784087 on Build GRCh38
rs192042969 Chr.9: 37784095-37784095 on Build GRCh38
rs771577074 Chr.9: 37783977-37783977 on Build GRCh38
rs753042657 Chr.9: 37783960-37783960 on Build GRCh38
rs201949026 Chr.9: 37784089-37784089 on Build GRCh38

View Full Product Details