Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89311184-89311429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKS2
Entrez Gene ID
1164
Gene Name
CDC28 protein kinase regulatory subunit 2
Gene Aliases
CKSHS2
UniGene
Hs.83758
Gene Chromosome Location
Chr.9: 89311195-89316703 on Build GRCh38
Transcripts
NM_001827.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6247312 c.33G>T p.K11N Chr.9: 89311325-89311325 on Build GRCh38
5795024 c.59+1G>C p.? Chr.9: 89311352-89311352 on Build GRCh38
5761629 c.59G>A p.R20Q Chr.9: 89311351-89311351 on Build GRCh38
6576230 c.20A>C p.Y7S Chr.9: 89311312-89311312 on Build GRCh38
6372825 c.13C>A p.Q5K Chr.9: 89311305-89311305 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758862660 Chr.9: 89311369-89311369 on Build GRCh38
rs374976316 Chr.9: 89311302-89311302 on Build GRCh38
rs984435346 Chr.9: 89311213-89311213 on Build GRCh38
rs746331942 Chr.9: 89311364-89311364 on Build GRCh38
rs780897091 Chr.9: 89311270-89311270 on Build GRCh38
rs551117215 Chr.9: 89311279-89311279 on Build GRCh38
rs926367637 Chr.9: 89311236-89311236 on Build GRCh38
rs1052637276 Chr.9: 89311312-89311312 on Build GRCh38
rs774800361 Chr.9: 89311251-89311251 on Build GRCh38
rs371951676 Chr.9: 89311288-89311288 on Build GRCh38
rs148627013 Chr.9: 89311210-89311210 on Build GRCh38
rs541610603 Chr.9: 89311216-89311216 on Build GRCh38
rs555808947 Chr.9: 89311377-89311377 on Build GRCh38
rs777304907 Chr.9: 89311257-89311257 on Build GRCh38
rs767367446 Chr.9: 89311390-89311390 on Build GRCh38
rs374633083 Chr.9: 89311243-89311243 on Build GRCh38
rs984570643 Chr.9: 89311233-89311233 on Build GRCh38
rs992251709 Chr.9: 89311239-89311239 on Build GRCh38
rs1035315861 Chr.9: 89311230-89311230 on Build GRCh38
rs768767950 Chr.9: 89311299-89311299 on Build GRCh38
rs893934113 Chr.9: 89311219-89311219 on Build GRCh38
rs945644670 Chr.9: 89311246-89311246 on Build GRCh38
rs747794592 Chr.9: 89311247-89311247 on Build GRCh38
rs572744073 Chr.9: 89311392-89311392 on Build GRCh38
rs905170424 Chr.9: 89311227-89311227 on Build GRCh38
rs1056467693 Chr.9: 89311393-89311393 on Build GRCh38
rs774249325 Chr.9: 89311308-89311308 on Build GRCh38
rs752344927 Chr.9: 89311272-89311272 on Build GRCh38
rs748415585 Chr.9: 89311380-89311380 on Build GRCh38
rs141662758 Chr.9: 89311319-89311319 on Build GRCh38
rs374151852 Chr.9: 89311213-89311213 on Build GRCh38
rs760914609 Chr.9: 89311298-89311298 on Build GRCh38
rs901739509 Chr.9: 89311292-89311292 on Build GRCh38
rs761954896 Chr.9: 89311217-89311217 on Build GRCh38
rs937620429 Chr.9: 89311237-89311237 on Build GRCh38
rs775744417 Chr.9: 89311293-89311293 on Build GRCh38
rs543490628 Chr.9: 89311225-89311225 on Build GRCh38
rs774566263 Chr.9: 89311212-89311212 on Build GRCh38
rs769815844 Chr.9: 89311381-89311381 on Build GRCh38
rs759450598 Chr.9: 89311389-89311389 on Build GRCh38
rs770624927 Chr.9: 89311286-89311286 on Build GRCh38
rs982621634 Chr.9: 89311241-89311241 on Build GRCh38
rs779558698 Chr.9: 89311287-89311287 on Build GRCh38
rs763642909 Chr.9: 89311349-89311349 on Build GRCh38
rs564186726 Chr.9: 89311218-89311218 on Build GRCh38
rs767587208 Chr.9: 89311234-89311234 on Build GRCh38
rs576723541 Chr.9: 89311204-89311204 on Build GRCh38
rs548983921 Chr.9: 89311262-89311262 on Build GRCh38
rs1010987730 Chr.9: 89311223-89311223 on Build GRCh38
rs182461066 Chr.9: 89311358-89311358 on Build GRCh38
rs935594172 Chr.9: 89311304-89311304 on Build GRCh38
rs536177306 Chr.9: 89311373-89311373 on Build GRCh38
rs755751583 Chr.9: 89311274-89311274 on Build GRCh38
rs753520463 Chr.9: 89311402-89311402 on Build GRCh38
rs755734988 Chr.9: 89311335-89311335 on Build GRCh38
rs775557912 Chr.9: 89311397-89311397 on Build GRCh38
rs766287968 Chr.9: 89311263-89311263 on Build GRCh38
rs984625657 Chr.9: 89311338-89311338 on Build GRCh38
rs764021275 Chr.9: 89311400-89311400 on Build GRCh38
rs147080616 Chr.9: 89311331-89311331 on Build GRCh38
rs3211664 Chr.9: 89311205-89311205 on Build GRCh38
rs563127195 Chr.9: 89311228-89311228 on Build GRCh38
rs754822026 Chr.9: 89311244-89311244 on Build GRCh38
rs760385286 Chr.9: 89311399-89311399 on Build GRCh38
rs1028272809 Chr.9: 89311224-89311224 on Build GRCh38
rs570613124 Chr.9: 89311301-89311301 on Build GRCh38
rs368569573 Chr.9: 89311248-89311248 on Build GRCh38
rs1026137627 Chr.9: 89311325-89311325 on Build GRCh38
rs756996469 Chr.9: 89311403-89311403 on Build GRCh38
rs759966109 Chr.9: 89311252-89311252 on Build GRCh38
rs747318666 Chr.9: 89311372-89311372 on Build GRCh38
rs3211665 Chr.9: 89311209-89311209 on Build GRCh38
rs370114711 Chr.9: 89311242-89311242 on Build GRCh38
rs369874363 Chr.9: 89311361-89311361 on Build GRCh38
rs11548887 Chr.9: 89311265-89311265 on Build GRCh38
rs11137560 Chr.9: 89311385-89311385 on Build GRCh38
rs775754220 Chr.9: 89311256-89311256 on Build GRCh38
rs951770925 Chr.9: 89311231-89311231 on Build GRCh38
rs1020063885 Chr.9: 89311281-89311281 on Build GRCh38
rs771571195 Chr.9: 89311249-89311249 on Build GRCh38
rs368906806 Chr.9: 89311404-89311404 on Build GRCh38
rs533019243 Chr.9: 89311222-89311222 on Build GRCh38
rs372028197 Chr.9: 89311258-89311258 on Build GRCh38
rs200458872 Chr.9: 89311370-89311370 on Build GRCh38
rs772385694 Chr.9: 89311253-89311253 on Build GRCh38
rs765337849 Chr.9: 89311305-89311305 on Build GRCh38
rs780889397 Chr.9: 89311245-89311245 on Build GRCh38
rs748821647 Chr.9: 89311284-89311284 on Build GRCh38
rs368132606 Chr.9: 89311269-89311269 on Build GRCh38
rs761873716 Chr.9: 89311303-89311303 on Build GRCh38

View Full Product Details