Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40987289-40987558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3665008 c.403G>A p.E135K Chr.1: 40987437-40987437 on Build GRCh38
1667476 c.347A>T p.H116L Chr.1: 40987381-40987381 on Build GRCh38
4238155 c.398T>C p.M133T Chr.1: 40987411-40987411 on Build GRCh38
6063548 c.385G>T p.A129S Chr.1: 40987419-40987419 on Build GRCh38
6063549 c.406G>T p.A136S Chr.1: 40987419-40987419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772244024 Chr.1: 40987448-40987448 on Build GRCh38
rs199692447 Chr.1: 40987514-40987514 on Build GRCh38
rs143058120 Chr.1: 40987420-40987420 on Build GRCh38
rs779871759 Chr.1: 40987372-40987372 on Build GRCh38
rs944805672 Chr.1: 40987528-40987528 on Build GRCh38
rs762909135 Chr.1: 40987333-40987333 on Build GRCh38
rs775731325 Chr.1: 40987457-40987457 on Build GRCh38
rs758062777 Chr.1: 40987367-40987367 on Build GRCh38
rs759635738 Chr.1: 40987421-40987421 on Build GRCh38
rs767739876 Chr.1: 40987341-40987341 on Build GRCh38
rs181188977 Chr.1: 40987496-40987496 on Build GRCh38
rs753719232 Chr.1: 40987492-40987492 on Build GRCh38
rs372708430 Chr.1: 40987502-40987502 on Build GRCh38
rs770231918 Chr.1: 40987385-40987385 on Build GRCh38
rs778555647 Chr.1: 40987356-40987356 on Build GRCh38
rs756711000 Chr.1: 40987355-40987355 on Build GRCh38
rs749862542 Chr.1: 40987403-40987403 on Build GRCh38
rs748480280 Chr.1: 40987376-40987376 on Build GRCh38
rs766528590 Chr.1: 40987338-40987338 on Build GRCh38
rs76891977 Chr.1: 40987459-40987459 on Build GRCh38
rs773797686 Chr.1: 40987332-40987332 on Build GRCh38
rs35368773 Chr.1: 40987350-40987351 on Build GRCh38
rs765095494 Chr.1: 40987499-40987499 on Build GRCh38
rs749962070 Chr.1: 40987361-40987361 on Build GRCh38
rs190246847 Chr.1: 40987321-40987321 on Build GRCh38
rs760367234 Chr.1: 40987468-40987468 on Build GRCh38
rs200274195 Chr.1: 40987395-40987395 on Build GRCh38
rs763909124 Chr.1: 40987349-40987349 on Build GRCh38
rs369068887 Chr.1: 40987414-40987414 on Build GRCh38
rs764010545 Chr.1: 40987486-40987486 on Build GRCh38
rs770147683 Chr.1: 40987325-40987325 on Build GRCh38
rs759821555 Chr.1: 40987340-40987340 on Build GRCh38
rs770911785 Chr.1: 40987416-40987416 on Build GRCh38
rs150745451 Chr.1: 40987339-40987339 on Build GRCh38
rs969931235 Chr.1: 40987407-40987407 on Build GRCh38
rs1052905715 Chr.1: 40987536-40987536 on Build GRCh38
rs753864692 Chr.1: 40987351-40987351 on Build GRCh38
rs758009873 Chr.1: 40987504-40987504 on Build GRCh38
rs980051840 Chr.1: 40987410-40987410 on Build GRCh38

View Full Product Details