Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37009221-37009490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC8
Entrez Gene ID
11340
Gene Name
exosome component 8
Gene Aliases
CIP3, EAP2, OIP2, PCH1C, RRP43, Rrp43p, bA421P11.3, p9
UniGene
Hs.294041
Gene Chromosome Location
Chr.13: 37000373-37009614 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1 NM_001014286.2 NM_001278480.1 NM_001278481.1 NM_001278482.1 NM_017569.3 XM_005266447.2 XM_005266449.3 XM_005266451.3 XM_005266454.3 XM_005266455.2 XM_005266456.3 XM_005266457.3 XM_005266458.3 XM_005266460.2 XM_005266461.3 XM_005266462.4 XM_005266464.3 XM_005266465.2 XM_005266467.3 XM_017020653.1 XM_017020654.1 XM_017020655.1 XM_017020656.1 XM_017020657.1 XM_017020658.1 XM_017020659.1 XM_017020660.1 XM_017020661.1

Gene Symbol
SUPT20H
Entrez Gene ID
55578
Gene Name
SPT20 homolog, SAGA complex component
Gene Aliases
C13, C13orf19, FAM48A, FP757, P38IP, SPT20
UniGene
Hs.435815
Gene Chromosome Location
Chr.13: 37009312-37059714 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1 NM_001014286.2 NM_001278480.1 NM_001278481.1 NM_001278482.1 NM_017569.3 XM_005266447.2 XM_005266449.3 XM_005266451.3 XM_005266454.3 XM_005266455.2 XM_005266456.3 XM_005266457.3 XM_005266458.3 XM_005266460.2 XM_005266461.3 XM_005266462.4 XM_005266464.3 XM_005266465.2 XM_005266467.3 XM_017020653.1 XM_017020654.1 XM_017020655.1 XM_017020656.1 XM_017020657.1 XM_017020658.1 XM_017020659.1 XM_017020660.1 XM_017020661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1366713 c.760C>T p.R254* Chr.13: 37009228-37009228 on Build GRCh38
3987382 c.782A>G p.E261G Chr.13: 37009250-37009250 on Build GRCh38
1194515 c.781G>C p.E261Q Chr.13: 37009249-37009249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745323976 Chr.13: 37009248-37009248 on Build GRCh38
rs573868857 Chr.13: 37009348-37009348 on Build GRCh38
rs766967892 Chr.13: 37009248-37009249 on Build GRCh38
rs117340034 Chr.13: 37009341-37009341 on Build GRCh38
rs966678435 Chr.13: 37009337-37009337 on Build GRCh38
rs36027220 Chr.13: 37009283-37009283 on Build GRCh38
rs765445348 Chr.13: 37009285-37009285 on Build GRCh38
rs759591829 Chr.13: 37009304-37009306 on Build GRCh38
rs767303775 Chr.13: 37009276-37009276 on Build GRCh38
rs750405455 Chr.13: 37009282-37009282 on Build GRCh38
rs776552289 Chr.13: 37009263-37009263 on Build GRCh38
rs752915989 Chr.13: 37009351-37009352 on Build GRCh38
rs1044235572 Chr.13: 37009297-37009297 on Build GRCh38
rs570100301 Chr.13: 37009307-37009307 on Build GRCh38
rs537421331 Chr.13: 37009335-37009335 on Build GRCh38
rs748969028 Chr.13: 37009351-37009351 on Build GRCh38
rs775092987 Chr.13: 37009254-37009254 on Build GRCh38
rs375388081 Chr.13: 37009270-37009270 on Build GRCh38
rs915746741 Chr.13: 37009290-37009290 on Build GRCh38
rs769326647 Chr.13: 37009250-37009250 on Build GRCh38
rs767735943 Chr.13: 37009329-37009330 on Build GRCh38
rs147730396 Chr.13: 37009255-37009255 on Build GRCh38
rs752942652 Chr.13: 37009295-37009295 on Build GRCh38
rs934339019 Chr.13: 37009466-37009466 on Build GRCh38
rs746414616 Chr.13: 37009339-37009339 on Build GRCh38
rs369711913 Chr.13: 37009311-37009311 on Build GRCh38
rs758605089 Chr.13: 37009301-37009301 on Build GRCh38
rs867831995 Chr.13: 37009266-37009266 on Build GRCh38
rs763339363 Chr.13: 37009246-37009249 on Build GRCh38
rs377709055 Chr.13: 37009361-37009361 on Build GRCh38
rs924299547 Chr.13: 37009382-37009382 on Build GRCh38
rs768485487 Chr.13: 37009261-37009261 on Build GRCh38
rs540927550 Chr.13: 37009431-37009431 on Build GRCh38
rs757653131 Chr.13: 37009321-37009321 on Build GRCh38
rs752021211 Chr.13: 37009255-37009257 on Build GRCh38
rs990684783 Chr.13: 37009381-37009381 on Build GRCh38

View Full Product Details