Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 98884919-98885154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E
Entrez Gene ID
1977
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E
UniGene
Hs.249718
Gene Chromosome Location
Chr.4: 98878456-98930635 on Build GRCh38
Transcripts
NM_001130678.1 NM_001130679.1 NM_001968.3 XM_006714126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680641 c.531A>G p.T177T Chr.4: 98884930-98884930 on Build GRCh38
6535393 c.418G>C p.E140Q Chr.4: 98885043-98885043 on Build GRCh38
6545634 c.481G>A p.D161N Chr.4: 98884980-98884980 on Build GRCh38
35428 c.453C>T p.G151G Chr.4: 98885008-98885008 on Build GRCh38
6652925 c.402T>C p.L134L Chr.4: 98885059-98885059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765013808 Chr.4: 98885099-98885099 on Build GRCh38
rs773473491 Chr.4: 98885067-98885067 on Build GRCh38
rs753775224 Chr.4: 98885093-98885093 on Build GRCh38
rs185066219 Chr.4: 98885008-98885008 on Build GRCh38
rs760805893 Chr.4: 98885070-98885070 on Build GRCh38
rs372999306 Chr.4: 98884966-98884966 on Build GRCh38
rs767620602 Chr.4: 98885065-98885065 on Build GRCh38
rs766446576 Chr.4: 98885088-98885088 on Build GRCh38
rs968700652 Chr.4: 98885016-98885016 on Build GRCh38
rs758129582 Chr.4: 98885111-98885111 on Build GRCh38
rs762161932 Chr.4: 98885050-98885050 on Build GRCh38
rs910675000 Chr.4: 98885109-98885109 on Build GRCh38
rs777386723 Chr.4: 98885112-98885112 on Build GRCh38
rs774451037 Chr.4: 98885020-98885020 on Build GRCh38
rs752434060 Chr.4: 98885105-98885105 on Build GRCh38
rs760614564 Chr.4: 98885064-98885065 on Build GRCh38
rs114497468 Chr.4: 98885095-98885095 on Build GRCh38
rs749648246 Chr.4: 98884989-98884989 on Build GRCh38
rs775645971 Chr.4: 98885065-98885065 on Build GRCh38
rs746565912 Chr.4: 98885113-98885113 on Build GRCh38
rs1004666356 Chr.4: 98885091-98885091 on Build GRCh38

View Full Product Details