Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55964267-55964496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP135
Entrez Gene ID
9662
Gene Name
centrosomal protein 135
Gene Aliases
CEP4, KIAA0635, MCPH8
UniGene
Hs.518767
Gene Chromosome Location
Chr.4: 55948808-56033363 on Build GRCh38
Transcripts
NM_025009.4 XM_005265788.3 XM_006714055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3301806 c.759C>T p.S253S Chr.4: 55964333-55964333 on Build GRCh38
288319 c.715C>T p.R239* Chr.4: 55964289-55964289 on Build GRCh38
1725720 c.748G>C p.G250R Chr.4: 55964322-55964322 on Build GRCh38
6100246 c.717A>C p.R239R Chr.4: 55964291-55964291 on Build GRCh38
87745 c.809T>C p.I270T Chr.4: 55964383-55964383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775520460 Chr.4: 55964347-55964347 on Build GRCh38
rs546711598 Chr.4: 55964414-55964414 on Build GRCh38
rs910957841 Chr.4: 55964461-55964461 on Build GRCh38
rs140245520 Chr.4: 55964368-55964368 on Build GRCh38
rs751061097 Chr.4: 55964443-55964443 on Build GRCh38
rs745647689 Chr.4: 55964340-55964340 on Build GRCh38
rs778315177 Chr.4: 55964442-55964442 on Build GRCh38
rs2593070 Chr.4: 55964455-55964455 on Build GRCh38
rs746592710 Chr.4: 55964325-55964325 on Build GRCh38
rs762997285 Chr.4: 55964362-55964362 on Build GRCh38
rs369023405 Chr.4: 55964409-55964409 on Build GRCh38
rs62308567 Chr.4: 55964439-55964439 on Build GRCh38
rs111958646 Chr.4: 55964387-55964387 on Build GRCh38
rs980201124 Chr.4: 55964326-55964326 on Build GRCh38
rs551280360 Chr.4: 55964382-55964382 on Build GRCh38
rs780858722 Chr.4: 55964449-55964449 on Build GRCh38
rs372973005 Chr.4: 55964296-55964296 on Build GRCh38
rs756760108 Chr.4: 55964448-55964448 on Build GRCh38
rs753230629 Chr.4: 55964308-55964308 on Build GRCh38
rs777274487 Chr.4: 55964321-55964321 on Build GRCh38
rs753142287 Chr.4: 55964428-55964428 on Build GRCh38
rs757851798 Chr.4: 55964309-55964309 on Build GRCh38
rs770607966 Chr.4: 55964328-55964328 on Build GRCh38
rs371746032 Chr.4: 55964314-55964314 on Build GRCh38
rs755345091 Chr.4: 55964297-55964297 on Build GRCh38
rs200221082 Chr.4: 55964301-55964301 on Build GRCh38
rs780760034 Chr.4: 55964329-55964329 on Build GRCh38
rs754044070 Chr.4: 55964395-55964395 on Build GRCh38
rs774438328 Chr.4: 55964411-55964411 on Build GRCh38
rs760723459 Chr.4: 55964371-55964371 on Build GRCh38
rs769509715 Chr.4: 55964342-55964342 on Build GRCh38
rs1035164786 Chr.4: 55964352-55964352 on Build GRCh38
rs962905274 Chr.4: 55964360-55964360 on Build GRCh38
rs2702355 Chr.4: 55964367-55964367 on Build GRCh38

View Full Product Details