Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9704542-9704767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE9
Entrez Gene ID
151835
Gene Name
copine family member 9
Gene Aliases
-
UniGene
Hs.121335
Gene Chromosome Location
Chr.3: 9703807-9729908 on Build GRCh38
Transcripts
NM_001308388.1 NM_153635.2 XM_011533386.2 XM_011533388.2 XM_011533389.2 XM_017005747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674408 c.71A>G p.N24S Chr.3: 9704589-9704589 on Build GRCh38
4989414 c.115G>C p.V39L Chr.3: 9704754-9704754 on Build GRCh38
4989415 c.115G>C p.V39L Chr.3: 9704754-9704754 on Build GRCh38
356976 c.85G>C p.D29H Chr.3: 9704603-9704603 on Build GRCh38
4674407 c.71A>G p.N24S Chr.3: 9704589-9704589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201088175 Chr.3: 9704572-9704572 on Build GRCh38
rs1049745426 Chr.3: 9704680-9704680 on Build GRCh38
rs901191640 Chr.3: 9704687-9704687 on Build GRCh38
rs1053600189 Chr.3: 9704734-9704734 on Build GRCh38
rs772992297 Chr.3: 9704702-9704702 on Build GRCh38
rs943997612 Chr.3: 9704698-9704698 on Build GRCh38
rs778296398 Chr.3: 9704656-9704656 on Build GRCh38
rs1032361546 Chr.3: 9704647-9704647 on Build GRCh38
rs879011954 Chr.3: 9704705-9704705 on Build GRCh38
rs980938286 Chr.3: 9704711-9704711 on Build GRCh38
rs908751593 Chr.3: 9704635-9704635 on Build GRCh38
rs752477274 Chr.3: 9704623-9704623 on Build GRCh38
rs536053391 Chr.3: 9704653-9704653 on Build GRCh38
rs554787788 Chr.3: 9704671-9704671 on Build GRCh38
rs767695683 Chr.3: 9704720-9704720 on Build GRCh38
rs201553272 Chr.3: 9704713-9704713 on Build GRCh38
rs774872743 Chr.3: 9704666-9704666 on Build GRCh38
rs756393532 Chr.3: 9704655-9704655 on Build GRCh38
rs750202521 Chr.3: 9704584-9704584 on Build GRCh38
rs116396781 Chr.3: 9704688-9704688 on Build GRCh38
rs1025036515 Chr.3: 9704682-9704682 on Build GRCh38
rs558442977 Chr.3: 9704704-9704704 on Build GRCh38
rs984752291 Chr.3: 9704740-9704740 on Build GRCh38
rs866778091 Chr.3: 9704576-9704576 on Build GRCh38
rs756409834 Chr.3: 9704741-9704741 on Build GRCh38
rs775997833 Chr.3: 9704675-9704675 on Build GRCh38
rs571231612 Chr.3: 9704564-9704564 on Build GRCh38
rs766303858 Chr.3: 9704620-9704620 on Build GRCh38
rs550679285 Chr.3: 9704607-9704607 on Build GRCh38
rs199726750 Chr.3: 9704703-9704703 on Build GRCh38
rs879137587 Chr.3: 9704700-9704700 on Build GRCh38
rs200234448 Chr.3: 9704650-9704650 on Build GRCh38
rs200458172 Chr.3: 9704658-9704658 on Build GRCh38
rs752768911 Chr.3: 9704730-9704730 on Build GRCh38
rs751618102 Chr.3: 9704626-9704626 on Build GRCh38
rs377066303 Chr.3: 9704701-9704701 on Build GRCh38
rs746337378 Chr.3: 9704669-9704669 on Build GRCh38

View Full Product Details