Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109252654-109252881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
UniGene
Hs.57652
Gene Chromosome Location
Chr.1: 109249705-109275756 on Build GRCh38
Transcripts
NM_001408.2 XM_005270580.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5457651 c.2624C>T p.T875M Chr.1: 109252703-109252703 on Build GRCh38
6873479 c.2779G>A p.D927N Chr.1: 109252858-109252858 on Build GRCh38
4482423 c.2603C>T p.S868F Chr.1: 109252682-109252682 on Build GRCh38
6448277 c.2621G>A p.R874Q Chr.1: 109252700-109252700 on Build GRCh38
200917 c.2705G>A p.R902H Chr.1: 109252784-109252784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367691599 Chr.1: 109252729-109252729 on Build GRCh38
rs753105253 Chr.1: 109252812-109252812 on Build GRCh38
rs745566729 Chr.1: 109252685-109252685 on Build GRCh38
rs776946440 Chr.1: 109252726-109252726 on Build GRCh38
rs770965352 Chr.1: 109252709-109252709 on Build GRCh38
rs772622829 Chr.1: 109252703-109252703 on Build GRCh38
rs748599721 Chr.1: 109252698-109252698 on Build GRCh38
rs368556615 Chr.1: 109252847-109252847 on Build GRCh38
rs138887323 Chr.1: 109252708-109252708 on Build GRCh38
rs371935118 Chr.1: 109252792-109252792 on Build GRCh38
rs766127459 Chr.1: 109252705-109252705 on Build GRCh38
rs775438398 Chr.1: 109252741-109252741 on Build GRCh38
rs374928321 Chr.1: 109252707-109252707 on Build GRCh38
rs779256212 Chr.1: 109252695-109252695 on Build GRCh38
rs779744418 Chr.1: 109252810-109252810 on Build GRCh38
rs375909210 Chr.1: 109252808-109252808 on Build GRCh38
rs759867572 Chr.1: 109252728-109252728 on Build GRCh38
rs747494318 Chr.1: 109252843-109252843 on Build GRCh38
rs758729822 Chr.1: 109252836-109252836 on Build GRCh38
rs962268647 Chr.1: 109252704-109252704 on Build GRCh38
rs201437569 Chr.1: 109252827-109252827 on Build GRCh38
rs529873677 Chr.1: 109252735-109252735 on Build GRCh38
rs987828949 Chr.1: 109252747-109252747 on Build GRCh38
rs761361328 Chr.1: 109252779-109252779 on Build GRCh38
rs751488741 Chr.1: 109252761-109252761 on Build GRCh38
rs150322062 Chr.1: 109252720-109252720 on Build GRCh38
rs774201169 Chr.1: 109252680-109252681 on Build GRCh38
rs915968408 Chr.1: 109252775-109252775 on Build GRCh38
rs1006432764 Chr.1: 109252680-109252680 on Build GRCh38
rs138048818 Chr.1: 109252748-109252748 on Build GRCh38
rs199806460 Chr.1: 109252787-109252787 on Build GRCh38
rs762667468 Chr.1: 109252686-109252686 on Build GRCh38
rs778322362 Chr.1: 109252842-109252842 on Build GRCh38
rs759318308 Chr.1: 109252765-109252765 on Build GRCh38
rs767034475 Chr.1: 109252781-109252781 on Build GRCh38
rs764084072 Chr.1: 109252752-109252752 on Build GRCh38

View Full Product Details