Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58531767-58532040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT1
Entrez Gene ID
23019
Gene Name
CCR4-NOT transcription complex subunit 1
Gene Aliases
AD-005, CDC39, NOT1, NOT1H
UniGene
Hs.460923
Gene Chromosome Location
Chr.16: 58519946-58629886 on Build GRCh38
Transcripts
NM_001265612.1 NM_016284.4 NR_049763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3969621 c.6164C>T p.T2055M Chr.16: 58531971-58531971 on Build GRCh38
972005 c.6133C>T p.R2045W Chr.16: 58532002-58532002 on Build GRCh38
6631368 c.6175_6177delAAG p.K2059delK Chr.16: 58531958-58531960 on Build GRCh38
6365169 c.6163A>G p.T2055A Chr.16: 58531972-58531972 on Build GRCh38
7134005 c.6135G>A p.R2045R Chr.16: 58532000-58532000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558865392 Chr.16: 58531923-58531923 on Build GRCh38
rs759871958 Chr.16: 58531807-58531807 on Build GRCh38
rs780957234 Chr.16: 58531992-58531992 on Build GRCh38
rs980020819 Chr.16: 58531877-58531877 on Build GRCh38
rs921159612 Chr.16: 58531889-58531889 on Build GRCh38
rs779159730 Chr.16: 58531967-58531967 on Build GRCh38
rs192664865 Chr.16: 58531890-58531890 on Build GRCh38
rs987974929 Chr.16: 58531790-58531790 on Build GRCh38
rs761242702 Chr.16: 58531912-58531912 on Build GRCh38
rs201226960 Chr.16: 58531970-58531970 on Build GRCh38
rs773567673 Chr.16: 58531916-58531919 on Build GRCh38
rs750894076 Chr.16: 58531908-58531908 on Build GRCh38
rs755054900 Chr.16: 58531924-58531924 on Build GRCh38
rs770095338 Chr.16: 58531879-58531879 on Build GRCh38
rs766854370 Chr.16: 58531922-58531922 on Build GRCh38
rs368992562 Chr.16: 58531855-58531855 on Build GRCh38
rs968380110 Chr.16: 58531814-58531814 on Build GRCh38
rs540057529 Chr.16: 58531858-58531858 on Build GRCh38
rs745718292 Chr.16: 58532008-58532008 on Build GRCh38
rs573220072 Chr.16: 58531813-58531816 on Build GRCh38
rs368784227 Chr.16: 58531937-58531937 on Build GRCh38
rs561571808 Chr.16: 58531860-58531860 on Build GRCh38
rs939751807 Chr.16: 58531947-58531947 on Build GRCh38

View Full Product Details