Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74671392-74671665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC3
Entrez Gene ID
80153
Gene Name
enhancer of mRNA decapping 3
Gene Aliases
LSM16, MRT50, YJDC, YJEFN2
UniGene
Hs.682454
Gene Chromosome Location
Chr.15: 74630558-74696045 on Build GRCh38
Transcripts
NM_001142443.1 NM_001142444.1 NM_025083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503955 c.439G>A p.E147K Chr.15: 74671500-74671500 on Build GRCh38
5383827 c.351C>T p.I117I Chr.15: 74671588-74671588 on Build GRCh38
6276048 c.285C>A p.I95I Chr.15: 74671654-74671654 on Build GRCh38
1708396 c.392C>T p.S131F Chr.15: 74671547-74671547 on Build GRCh38
4390173 c.365A>C p.D122A Chr.15: 74671574-74671574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762672355 Chr.15: 74671469-74671469 on Build GRCh38
rs767501602 Chr.15: 74671475-74671475 on Build GRCh38
rs759094705 Chr.15: 74671433-74671433 on Build GRCh38
rs779063604 Chr.15: 74671513-74671513 on Build GRCh38
rs777978304 Chr.15: 74671635-74671635 on Build GRCh38
rs1022854363 Chr.15: 74671574-74671574 on Build GRCh38
rs968108909 Chr.15: 74671586-74671586 on Build GRCh38
rs757001010 Chr.15: 74671606-74671606 on Build GRCh38
rs939926716 Chr.15: 74671603-74671604 on Build GRCh38
rs748562304 Chr.15: 74671515-74671515 on Build GRCh38
rs766482884 Chr.15: 74671580-74671580 on Build GRCh38
rs750261315 Chr.15: 74671488-74671488 on Build GRCh38
rs1009263319 Chr.15: 74671419-74671419 on Build GRCh38
rs376760239 Chr.15: 74671503-74671503 on Build GRCh38
rs113901995 Chr.15: 74671509-74671509 on Build GRCh38
rs778414721 Chr.15: 74671520-74671520 on Build GRCh38
rs779114659 Chr.15: 74671584-74671584 on Build GRCh38
rs747461655 Chr.15: 74671525-74671525 on Build GRCh38
rs756283540 Chr.15: 74671496-74671496 on Build GRCh38
rs753948126 Chr.15: 74671583-74671583 on Build GRCh38
rs377587628 Chr.15: 74671421-74671421 on Build GRCh38
rs148327581 Chr.15: 74671544-74671544 on Build GRCh38
rs758859891 Chr.15: 74671625-74671625 on Build GRCh38
rs760503949 Chr.15: 74671571-74671571 on Build GRCh38
rs780091375 Chr.15: 74671501-74671501 on Build GRCh38
rs750428769 Chr.15: 74671566-74671566 on Build GRCh38
rs370722720 Chr.15: 74671440-74671440 on Build GRCh38
rs1003838369 Chr.15: 74671466-74671466 on Build GRCh38
rs979152587 Chr.15: 74671473-74671473 on Build GRCh38
rs544178799 Chr.15: 74671472-74671472 on Build GRCh38
rs767211729 Chr.15: 74671552-74671552 on Build GRCh38
rs772261057 Chr.15: 74671518-74671518 on Build GRCh38
rs542158660 Chr.15: 74671551-74671551 on Build GRCh38
rs773815619 Chr.15: 74671547-74671547 on Build GRCh38
rs753606943 Chr.15: 74671582-74671583 on Build GRCh38
rs764023124 Chr.15: 74671470-74671470 on Build GRCh38
rs752863254 Chr.15: 74671598-74671598 on Build GRCh38
rs967809528 Chr.15: 74671448-74671448 on Build GRCh38
rs917872534 Chr.15: 74671589-74671589 on Build GRCh38
rs1007558701 Chr.15: 74671594-74671594 on Build GRCh38
rs200931517 Chr.15: 74671543-74671543 on Build GRCh38
rs761464346 Chr.15: 74671474-74671474 on Build GRCh38
rs767215558 Chr.15: 74671545-74671545 on Build GRCh38
rs775342225 Chr.15: 74671454-74671454 on Build GRCh38
rs373160392 Chr.15: 74671416-74671416 on Build GRCh38
rs769477794 Chr.15: 74671531-74671531 on Build GRCh38
rs755158567 Chr.15: 74671512-74671512 on Build GRCh38

View Full Product Details