Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171334430-171334661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO4
Entrez Gene ID
2329
Gene Name
flavin containing monooxygenase 4
Gene Aliases
FMO2
UniGene
Hs.386502
Gene Chromosome Location
Chr.1: 171314131-171342084 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2 XM_011509349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123200 c.853G>A p.D285N Chr.1: 171334436-171334436 on Build GRCh38
3689129 c.1032C>G p.L344L Chr.1: 171334615-171334615 on Build GRCh38
3984466 c.962T>C p.I321T Chr.1: 171334545-171334545 on Build GRCh38
677183 c.976T>G p.F326V Chr.1: 171334559-171334559 on Build GRCh38
5483371 c.876C>A p.L292L Chr.1: 171334459-171334459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3737925 Chr.1: 171334505-171334505 on Build GRCh38
rs774400664 Chr.1: 171334574-171334574 on Build GRCh38
rs768559319 Chr.1: 171334470-171334470 on Build GRCh38
rs573059844 Chr.1: 171334487-171334487 on Build GRCh38
rs771136767 Chr.1: 171334604-171334604 on Build GRCh38
rs773375083 Chr.1: 171334602-171334602 on Build GRCh38
rs756951706 Chr.1: 171334538-171334538 on Build GRCh38
rs760967741 Chr.1: 171334603-171334603 on Build GRCh38
rs138313509 Chr.1: 171334549-171334549 on Build GRCh38
rs748266769 Chr.1: 171334581-171334581 on Build GRCh38
rs768646263 Chr.1: 171334563-171334563 on Build GRCh38
rs139018321 Chr.1: 171334548-171334548 on Build GRCh38
rs1042767 Chr.1: 171334551-171334551 on Build GRCh38
rs537208861 Chr.1: 171334457-171334457 on Build GRCh38
rs149258005 Chr.1: 171334618-171334618 on Build GRCh38
rs759950250 Chr.1: 171334628-171334628 on Build GRCh38
rs545819580 Chr.1: 171334617-171334617 on Build GRCh38
rs61342270 Chr.1: 171334598-171334598 on Build GRCh38
rs756978964 Chr.1: 171334486-171334486 on Build GRCh38
rs768035106 Chr.1: 171334618-171334636 on Build GRCh38
rs931705009 Chr.1: 171334614-171334614 on Build GRCh38
rs774417198 Chr.1: 171334475-171334475 on Build GRCh38
rs369627919 Chr.1: 171334631-171334631 on Build GRCh38
rs767784790 Chr.1: 171334477-171334477 on Build GRCh38
rs773536271 Chr.1: 171334478-171334478 on Build GRCh38
rs754244016 Chr.1: 171334491-171334491 on Build GRCh38
rs763695770 Chr.1: 171334531-171334531 on Build GRCh38
rs142455994 Chr.1: 171334492-171334492 on Build GRCh38
rs751176716 Chr.1: 171334535-171334535 on Build GRCh38
rs781027440 Chr.1: 171334547-171334547 on Build GRCh38
rs981838876 Chr.1: 171334481-171334481 on Build GRCh38
rs779909399 Chr.1: 171334550-171334550 on Build GRCh38
rs867902199 Chr.1: 171334466-171334466 on Build GRCh38
rs762017210 Chr.1: 171334476-171334476 on Build GRCh38
rs201971967 Chr.1: 171334507-171334507 on Build GRCh38
rs1022438335 Chr.1: 171334493-171334493 on Build GRCh38

View Full Product Details