Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82584046-82584271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK2
Entrez Gene ID
3607
Gene Name
forkhead box K2
Gene Aliases
ILF, ILF-1, ILF1
UniGene
Hs.591140
Gene Chromosome Location
Chr.17: 82519718-82604607 on Build GRCh38
Transcripts
NM_004514.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523424 c.1213G>T p.E405* Chr.17: 82584122-82584122 on Build GRCh38
3523890 c.1167C>T p.T389T Chr.17: 82584076-82584076 on Build GRCh38
7146809 c.1176C>T p.S392S Chr.17: 82584085-82584085 on Build GRCh38
6432491 c.1225G>A p.A409T Chr.17: 82584134-82584134 on Build GRCh38
6562626 c.1245C>G p.V415V Chr.17: 82584154-82584154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759519151 Chr.17: 82584113-82584113 on Build GRCh38
rs752777895 Chr.17: 82584125-82584125 on Build GRCh38
rs774554828 Chr.17: 82584199-82584199 on Build GRCh38
rs762170087 Chr.17: 82584201-82584201 on Build GRCh38
rs375893720 Chr.17: 82584091-82584091 on Build GRCh38
rs775102088 Chr.17: 82584239-82584239 on Build GRCh38
rs372920071 Chr.17: 82584198-82584198 on Build GRCh38
rs777313246 Chr.17: 82584226-82584226 on Build GRCh38
rs764015593 Chr.17: 82584140-82584140 on Build GRCh38
rs534392014 Chr.17: 82584181-82584181 on Build GRCh38
rs143594098 Chr.17: 82584130-82584130 on Build GRCh38
rs368830698 Chr.17: 82584168-82584168 on Build GRCh38
rs756641516 Chr.17: 82584154-82584154 on Build GRCh38
rs754398900 Chr.17: 82584219-82584219 on Build GRCh38
rs756512375 Chr.17: 82584218-82584218 on Build GRCh38
rs868546131 Chr.17: 82584120-82584120 on Build GRCh38
rs781725381 Chr.17: 82584150-82584150 on Build GRCh38
rs750871743 Chr.17: 82584210-82584210 on Build GRCh38
rs956703567 Chr.17: 82584230-82584230 on Build GRCh38
rs968733567 Chr.17: 82584242-82584242 on Build GRCh38
rs746684608 Chr.17: 82584175-82584175 on Build GRCh38
rs147224723 Chr.17: 82584151-82584151 on Build GRCh38
rs757508510 Chr.17: 82584148-82584148 on Build GRCh38
rs777223222 Chr.17: 82584169-82584169 on Build GRCh38
rs765216482 Chr.17: 82584115-82584115 on Build GRCh38
rs757703270 Chr.17: 82584067-82584067 on Build GRCh38
rs530148354 Chr.17: 82584109-82584109 on Build GRCh38
rs549650969 Chr.17: 82584238-82584238 on Build GRCh38
rs778322283 Chr.17: 82584090-82584090 on Build GRCh38
rs746490556 Chr.17: 82584228-82584228 on Build GRCh38
rs371497720 Chr.17: 82584100-82584100 on Build GRCh38
rs138125214 Chr.17: 82584068-82584068 on Build GRCh38
rs59764629 Chr.17: 82584225-82584225 on Build GRCh38
rs770315753 Chr.17: 82584088-82584088 on Build GRCh38
rs769418037 Chr.17: 82584182-82584182 on Build GRCh38
rs767843181 Chr.17: 82584206-82584206 on Build GRCh38
rs948959583 Chr.17: 82584131-82584131 on Build GRCh38
rs1037736759 Chr.17: 82584149-82584149 on Build GRCh38
rs770742063 Chr.17: 82584110-82584110 on Build GRCh38
rs144073500 Chr.17: 82584103-82584103 on Build GRCh38
rs368819935 Chr.17: 82584097-82584097 on Build GRCh38
rs138960969 Chr.17: 82584184-82584184 on Build GRCh38
rs368015086 Chr.17: 82584106-82584106 on Build GRCh38
rs150409126 Chr.17: 82584167-82584167 on Build GRCh38
rs746403456 Chr.17: 82584076-82584076 on Build GRCh38
rs372041686 Chr.17: 82584112-82584112 on Build GRCh38
rs200219871 Chr.17: 82584236-82584236 on Build GRCh38
rs977847180 Chr.17: 82584247-82584247 on Build GRCh38
rs770547682 Chr.17: 82584179-82584179 on Build GRCh38
rs538510925 Chr.17: 82584235-82584235 on Build GRCh38

View Full Product Details