Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50338838-50339111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL1
Entrez Gene ID
8814
Gene Name
cyclin dependent kinase like 1
Gene Aliases
KKIALRE, P42
UniGene
Hs.280881
Gene Chromosome Location
Chr.14: 50329404-50397404 on Build GRCh38
Transcripts
NM_001282236.1 NM_004196.4 XM_005268157.2 XM_005268159.2 XM_005268160.4 XM_011537276.1 XM_017021729.1 XM_017021730.1 XM_017021731.1 XM_017021732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3770073 c.741+5G>C p.? Chr.14: 50338942-50338942 on Build GRCh38
6328150 c.727G>A p.D243N Chr.14: 50338961-50338961 on Build GRCh38
6873472 c.702G>T p.Q234H Chr.14: 50338986-50338986 on Build GRCh38
1200587 c.671C>A p.P224H Chr.14: 50339017-50339017 on Build GRCh38
1200589 c.708C>A p.F236L Chr.14: 50338980-50338980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201870526 Chr.14: 50338921-50338921 on Build GRCh38
rs370305372 Chr.14: 50338937-50338937 on Build GRCh38
rs931779967 Chr.14: 50339024-50339024 on Build GRCh38
rs751381790 Chr.14: 50338988-50338988 on Build GRCh38
rs763849645 Chr.14: 50338929-50338929 on Build GRCh38
rs201211299 Chr.14: 50338948-50338948 on Build GRCh38
rs764708646 Chr.14: 50338946-50338946 on Build GRCh38
rs372665114 Chr.14: 50338939-50338939 on Build GRCh38
rs769207625 Chr.14: 50339062-50339062 on Build GRCh38
rs371181719 Chr.14: 50339054-50339054 on Build GRCh38
rs941115113 Chr.14: 50339041-50339041 on Build GRCh38
rs193193846 Chr.14: 50338977-50338977 on Build GRCh38
rs757490884 Chr.14: 50339010-50339010 on Build GRCh38
rs775592264 Chr.14: 50338942-50338943 on Build GRCh38
rs532557422 Chr.14: 50339051-50339051 on Build GRCh38
rs184318768 Chr.14: 50339022-50339022 on Build GRCh38
rs991407023 Chr.14: 50338920-50338920 on Build GRCh38
rs745772988 Chr.14: 50339038-50339038 on Build GRCh38
rs776027247 Chr.14: 50338923-50338923 on Build GRCh38
rs929672829 Chr.14: 50339083-50339083 on Build GRCh38
rs760258972 Chr.14: 50338918-50338918 on Build GRCh38
rs760993904 Chr.14: 50339045-50339046 on Build GRCh38
rs1037976389 Chr.14: 50339052-50339052 on Build GRCh38
rs147406709 Chr.14: 50338953-50338953 on Build GRCh38
rs772104961 Chr.14: 50338923-50338924 on Build GRCh38
rs754712040 Chr.14: 50338992-50338992 on Build GRCh38
rs769798660 Chr.14: 50339058-50339059 on Build GRCh38
rs777120136 Chr.14: 50339071-50339071 on Build GRCh38
rs778908810 Chr.14: 50339019-50339019 on Build GRCh38
rs761072122 Chr.14: 50338924-50338924 on Build GRCh38

View Full Product Details