Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141528491-141528762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH1
Entrez Gene ID
1729
Gene Name
diaphanous related formin 1
Gene Aliases
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
UniGene
Hs.529451
Gene Chromosome Location
Chr.5: 141515021-141619055 on Build GRCh38
Transcripts
NM_001079812.2 NM_001314007.1 NM_005219.4 XM_011537572.2 XM_011537573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620683 c.3053C>T p.T1018M Chr.5: 141528548-141528548 on Build GRCh38
3208978 c.3018G>A p.K1006K Chr.5: 141528702-141528702 on Build GRCh38
6560728 c.3019-7C>G p.? Chr.5: 141528589-141528589 on Build GRCh38
7196593 c.3011T>A p.L1004H Chr.5: 141528709-141528709 on Build GRCh38
3373725 c.3096C>G p.P1032P Chr.5: 141528505-141528505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969163545 Chr.5: 141528713-141528713 on Build GRCh38
rs566236870 Chr.5: 141528524-141528524 on Build GRCh38
rs370089310 Chr.5: 141528604-141528604 on Build GRCh38
rs757756780 Chr.5: 141528710-141528710 on Build GRCh38
rs376220834 Chr.5: 141528551-141528551 on Build GRCh38
rs752924382 Chr.5: 141528563-141528563 on Build GRCh38
rs772774094 Chr.5: 141528654-141528654 on Build GRCh38
rs752977576 Chr.5: 141528675-141528675 on Build GRCh38
rs1004400733 Chr.5: 141528544-141528544 on Build GRCh38
rs774379327 Chr.5: 141528661-141528661 on Build GRCh38
rs555292453 Chr.5: 141528574-141528574 on Build GRCh38
rs535140857 Chr.5: 141528532-141528532 on Build GRCh38
rs762935116 Chr.5: 141528538-141528538 on Build GRCh38
rs757781927 Chr.5: 141528592-141528592 on Build GRCh38
rs992133469 Chr.5: 141528726-141528726 on Build GRCh38
rs776298359 Chr.5: 141528630-141528630 on Build GRCh38
rs767351715 Chr.5: 141528666-141528666 on Build GRCh38
rs1022451587 Chr.5: 141528627-141528627 on Build GRCh38
rs373275414 Chr.5: 141528550-141528550 on Build GRCh38
rs772380259 Chr.5: 141528608-141528608 on Build GRCh38
rs920298762 Chr.5: 141528638-141528638 on Build GRCh38
rs759433754 Chr.5: 141528665-141528665 on Build GRCh38
rs371366611 Chr.5: 141528553-141528553 on Build GRCh38
rs777847342 Chr.5: 141528582-141528582 on Build GRCh38
rs373518450 Chr.5: 141528652-141528652 on Build GRCh38
rs1040527951 Chr.5: 141528598-141528598 on Build GRCh38
rs779499759 Chr.5: 141528721-141528721 on Build GRCh38
rs996764181 Chr.5: 141528622-141528622 on Build GRCh38
rs773028163 Chr.5: 141528537-141528537 on Build GRCh38
rs753927669 Chr.5: 141528707-141528707 on Build GRCh38
rs750890653 Chr.5: 141528738-141528738 on Build GRCh38
rs1006618137 Chr.5: 141528578-141528578 on Build GRCh38
rs727502960 Chr.5: 141528547-141528547 on Build GRCh38
rs760933407 Chr.5: 141528683-141528683 on Build GRCh38
rs764150939 Chr.5: 141528694-141528694 on Build GRCh38
rs376427289 Chr.5: 141528659-141528659 on Build GRCh38
rs767871722 Chr.5: 141528555-141528555 on Build GRCh38
rs761336460 Chr.5: 141528526-141528526 on Build GRCh38
rs575426192 Chr.5: 141528594-141528594 on Build GRCh38
rs749822608 Chr.5: 141528590-141528590 on Build GRCh38

View Full Product Details