Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27101102-27101349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGREF1
Entrez Gene ID
10669
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
CGR11
UniGene
Hs.159525
Gene Chromosome Location
Chr.2: 27099353-27119127 on Build GRCh38
Transcripts
NM_001166239.1 NM_001166240.1 NM_001301324.1 NM_006569.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6627657 c.923T>C p.V308A Chr.2: 27101308-27101308 on Build GRCh38
5782452 c.863A>C p.D288A Chr.2: 27101317-27101317 on Build GRCh38
6157671 c.907C>A p.Q303K Chr.2: 27101324-27101324 on Build GRCh38
6542450 c.903C>A p.I301I Chr.2: 27101277-27101277 on Build GRCh38
6627658 c.872T>C p.V291A Chr.2: 27101308-27101308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541369615 Chr.2: 27101191-27101191 on Build GRCh38
rs910152774 Chr.2: 27101131-27101131 on Build GRCh38
rs777756158 Chr.2: 27101294-27101294 on Build GRCh38
rs761927756 Chr.2: 27101254-27101254 on Build GRCh38
rs776522100 Chr.2: 27101249-27101249 on Build GRCh38
rs770729823 Chr.2: 27101306-27101306 on Build GRCh38
rs750958789 Chr.2: 27101264-27101264 on Build GRCh38
rs774622620 Chr.2: 27101168-27101169 on Build GRCh38
rs576480522 Chr.2: 27101144-27101144 on Build GRCh38
rs1039925901 Chr.2: 27101165-27101165 on Build GRCh38
rs942907260 Chr.2: 27101136-27101136 on Build GRCh38
rs760893473 Chr.2: 27101224-27101235 on Build GRCh38
rs761048772 Chr.2: 27101175-27101175 on Build GRCh38
rs116418742 Chr.2: 27101169-27101169 on Build GRCh38
rs1012609370 Chr.2: 27101218-27101218 on Build GRCh38
rs775751586 Chr.2: 27101228-27101228 on Build GRCh38
rs373596029 Chr.2: 27101230-27101230 on Build GRCh38
rs1057396 Chr.2: 27101139-27101139 on Build GRCh38
rs1023553457 Chr.2: 27101221-27101221 on Build GRCh38
rs146521833 Chr.2: 27101172-27101172 on Build GRCh38
rs386644208 Chr.2: 27101168-27101169 on Build GRCh38
rs559606850 Chr.2: 27101200-27101200 on Build GRCh38
rs765880445 Chr.2: 27101262-27101262 on Build GRCh38
rs533954435 Chr.2: 27101135-27101135 on Build GRCh38
rs541426747 Chr.2: 27101277-27101277 on Build GRCh38
rs1054826802 Chr.2: 27101205-27101205 on Build GRCh38
rs531648483 Chr.2: 27101182-27101182 on Build GRCh38
rs749213945 Chr.2: 27101300-27101300 on Build GRCh38
rs902204109 Chr.2: 27101194-27101194 on Build GRCh38
rs778615710 Chr.2: 27101307-27101307 on Build GRCh38
rs1031945479 Chr.2: 27101203-27101203 on Build GRCh38
rs140862111 Chr.2: 27101243-27101243 on Build GRCh38
rs751260296 Chr.2: 27101283-27101283 on Build GRCh38
rs759964516 Chr.2: 27101251-27101252 on Build GRCh38
rs1041812819 Chr.2: 27101171-27101171 on Build GRCh38
rs780778707 Chr.2: 27101290-27101290 on Build GRCh38
rs755620105 Chr.2: 27101292-27101292 on Build GRCh38
rs898306548 Chr.2: 27101158-27101158 on Build GRCh38
rs1028530581 Chr.2: 27101231-27101231 on Build GRCh38
rs752199429 Chr.2: 27101291-27101291 on Build GRCh38
rs759927208 Chr.2: 27101210-27101210 on Build GRCh38
rs909504355 Chr.2: 27101324-27101324 on Build GRCh38
rs755483363 Chr.2: 27101286-27101286 on Build GRCh38
rs1057394 Chr.2: 27101168-27101168 on Build GRCh38

View Full Product Details