Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 153944433-153944706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT13
Entrez Gene ID
114805
Gene Name
polypeptide N-acetylgalactosaminyltransferase 13
Gene Aliases
GalNAc-T13
UniGene
Hs.470277
Gene Chromosome Location
Chr.2: 153871898-154454177 on Build GRCh38
Transcripts
NM_001301627.1 NM_052917.3 XM_011510537.2 XM_017003258.1 XM_017003259.1 XM_017003260.1 XM_017003261.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397171 c.138G>T p.L46F Chr.2: 153944635-153944635 on Build GRCh38
3568039 c.118A>T p.R40* Chr.2: 153944615-153944615 on Build GRCh38
3568034 c.45G>A p.L15L Chr.2: 153944542-153944542 on Build GRCh38
1494579 c.25G>C p.V9L Chr.2: 153944522-153944522 on Build GRCh38
3568043 c.140G>A p.R47K Chr.2: 153944637-153944637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145025659 Chr.2: 153944598-153944598 on Build GRCh38
rs376838127 Chr.2: 153944562-153944562 on Build GRCh38
rs774393674 Chr.2: 153944674-153944674 on Build GRCh38
rs779241418 Chr.2: 153944665-153944668 on Build GRCh38
rs763447790 Chr.2: 153944615-153944615 on Build GRCh38
rs936304619 Chr.2: 153944675-153944675 on Build GRCh38
rs752295859 Chr.2: 153944516-153944516 on Build GRCh38
rs762728936 Chr.2: 153944583-153944583 on Build GRCh38
rs773210639 Chr.2: 153944575-153944575 on Build GRCh38
rs772328017 Chr.2: 153944543-153944543 on Build GRCh38
rs987656717 Chr.2: 153944573-153944573 on Build GRCh38
rs757817210 Chr.2: 153944633-153944633 on Build GRCh38
rs751258487 Chr.2: 153944511-153944511 on Build GRCh38
rs373908895 Chr.2: 153944494-153944494 on Build GRCh38
rs1401518 Chr.2: 153944486-153944486 on Build GRCh38
rs747138875 Chr.2: 153944664-153944664 on Build GRCh38
rs766409416 Chr.2: 153944597-153944597 on Build GRCh38
rs368853298 Chr.2: 153944462-153944462 on Build GRCh38
rs369723516 Chr.2: 153944656-153944656 on Build GRCh38
rs759378533 Chr.2: 153944600-153944600 on Build GRCh38
rs760649021 Chr.2: 153944482-153944482 on Build GRCh38
rs779319343 Chr.2: 153944528-153944528 on Build GRCh38
rs995125825 Chr.2: 153944500-153944500 on Build GRCh38
rs535801705 Chr.2: 153944601-153944601 on Build GRCh38
rs754739183 Chr.2: 153944513-153944513 on Build GRCh38
rs72865049 Chr.2: 153944683-153944683 on Build GRCh38
rs772979116 Chr.2: 153944491-153944491 on Build GRCh38
rs190449206 Chr.2: 153944669-153944669 on Build GRCh38
rs778367534 Chr.2: 153944520-153944520 on Build GRCh38
rs778955885 Chr.2: 153944500-153944502 on Build GRCh38
rs944702288 Chr.2: 153944589-153944589 on Build GRCh38
rs186038567 Chr.2: 153944607-153944607 on Build GRCh38
rs752243974 Chr.2: 153944606-153944606 on Build GRCh38
rs750826926 Chr.2: 153944637-153944637 on Build GRCh38
rs767259675 Chr.2: 153944514-153944514 on Build GRCh38
rs749700926 Chr.2: 153944524-153944524 on Build GRCh38
rs775542419 Chr.2: 153944549-153944549 on Build GRCh38
rs376635440 Chr.2: 153944672-153944672 on Build GRCh38
rs181234314 Chr.2: 153944466-153944466 on Build GRCh38
rs538808548 Chr.2: 153944525-153944525 on Build GRCh38
rs755543535 Chr.2: 153944517-153944517 on Build GRCh38
rs747142582 Chr.2: 153944560-153944560 on Build GRCh38
rs371563167 Chr.2: 153944679-153944679 on Build GRCh38
rs868667232 Chr.2: 153944495-153944495 on Build GRCh38
rs983539360 Chr.2: 153944485-153944485 on Build GRCh38
rs200101064 Chr.2: 153944676-153944676 on Build GRCh38
rs950831995 Chr.2: 153944563-153944563 on Build GRCh38
rs759430075 Chr.2: 153944678-153944678 on Build GRCh38
rs757439470 Chr.2: 153944606-153944608 on Build GRCh38
rs768783530 Chr.2: 153944666-153944666 on Build GRCh38
rs779368773 Chr.2: 153944635-153944635 on Build GRCh38
rs572595038 Chr.2: 153944639-153944639 on Build GRCh38
rs1039063658 Chr.2: 153944602-153944602 on Build GRCh38
rs766217238 Chr.2: 153944507-153944507 on Build GRCh38

View Full Product Details