Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122834587-122834816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627356 c.1351G>C p.A451P Chr.10: 122834737-122834737 on Build GRCh38
4760994 c.1366G>T p.D456Y Chr.10: 122834722-122834722 on Build GRCh38
3435338 c.1374C>T p.I458I Chr.10: 122834714-122834714 on Build GRCh38
5578361 c.1315C>T p.L439F Chr.10: 122834773-122834773 on Build GRCh38
3414780 c.1373T>G p.I458S Chr.10: 122834715-122834715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562825865 Chr.10: 122834710-122834710 on Build GRCh38
rs972261730 Chr.10: 122834688-122834688 on Build GRCh38
rs140438956 Chr.10: 122834762-122834762 on Build GRCh38
rs531469000 Chr.10: 122834712-122834712 on Build GRCh38
rs756159212 Chr.10: 122834765-122834765 on Build GRCh38
rs917175051 Chr.10: 122834702-122834702 on Build GRCh38
rs763479860 Chr.10: 122834747-122834747 on Build GRCh38
rs960970216 Chr.10: 122834624-122834624 on Build GRCh38
rs757424430 Chr.10: 122834685-122834685 on Build GRCh38
rs778769903 Chr.10: 122834709-122834709 on Build GRCh38
rs781373762 Chr.10: 122834689-122834689 on Build GRCh38
rs375930865 Chr.10: 122834722-122834722 on Build GRCh38
rs368389572 Chr.10: 122834745-122834745 on Build GRCh38
rs563527324 Chr.10: 122834738-122834738 on Build GRCh38
rs773099334 Chr.10: 122834714-122834714 on Build GRCh38
rs199934875 Chr.10: 122834783-122834783 on Build GRCh38
rs758705118 Chr.10: 122834670-122834670 on Build GRCh38
rs370233601 Chr.10: 122834786-122834786 on Build GRCh38
rs750641520 Chr.10: 122834764-122834764 on Build GRCh38
rs772088116 Chr.10: 122834775-122834775 on Build GRCh38
rs962214756 Chr.10: 122834651-122834651 on Build GRCh38
rs527431751 Chr.10: 122834757-122834757 on Build GRCh38
rs370622556 Chr.10: 122834766-122834766 on Build GRCh38
rs748068980 Chr.10: 122834773-122834773 on Build GRCh38
rs751972937 Chr.10: 122834751-122834751 on Build GRCh38
rs148306521 Chr.10: 122834723-122834723 on Build GRCh38
rs368644334 Chr.10: 122834719-122834719 on Build GRCh38
rs372927471 Chr.10: 122834782-122834782 on Build GRCh38
rs758327968 Chr.10: 122834671-122834671 on Build GRCh38
rs909710199 Chr.10: 122834743-122834743 on Build GRCh38
rs11248321 Chr.10: 122834637-122834637 on Build GRCh38
rs769908981 Chr.10: 122834699-122834699 on Build GRCh38
rs751813445 Chr.10: 122834679-122834679 on Build GRCh38
rs571088257 Chr.10: 122834729-122834729 on Build GRCh38
rs745979723 Chr.10: 122834693-122834693 on Build GRCh38
rs764468170 Chr.10: 122834750-122834750 on Build GRCh38
rs985299572 Chr.10: 122834632-122834632 on Build GRCh38

View Full Product Details