Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29860502-29860731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDIPT
Entrez Gene ID
10423
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
PIS, PIS1
UniGene
Hs.121549
Gene Chromosome Location
Chr.16: 29858356-29863288 on Build GRCh38
Transcripts
NM_001286585.1 NM_001286586.1 NM_006319.4 XM_005255038.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545496 c.379G>A p.G127R Chr.16: 29860616-29860616 on Build GRCh38
7412874 c.414G>A p.R138R Chr.16: 29860581-29860581 on Build GRCh38
969672 c.379G>T p.G127W Chr.16: 29860616-29860616 on Build GRCh38
6625298 c.370G>A p.D124N Chr.16: 29860625-29860625 on Build GRCh38
4671065 c.369C>T p.I123I Chr.16: 29860626-29860626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557300967 Chr.16: 29860541-29860541 on Build GRCh38
rs779056289 Chr.16: 29860615-29860615 on Build GRCh38
rs754330645 Chr.16: 29860684-29860684 on Build GRCh38
rs757373468 Chr.16: 29860610-29860610 on Build GRCh38
rs750555293 Chr.16: 29860616-29860616 on Build GRCh38
rs767714724 Chr.16: 29860599-29860599 on Build GRCh38
rs754418666 Chr.16: 29860608-29860608 on Build GRCh38
rs558173049 Chr.16: 29860654-29860654 on Build GRCh38
rs764585113 Chr.16: 29860680-29860680 on Build GRCh38
rs765220084 Chr.16: 29860699-29860699 on Build GRCh38
rs368061228 Chr.16: 29860624-29860624 on Build GRCh38
rs891907273 Chr.16: 29860527-29860527 on Build GRCh38
rs761170255 Chr.16: 29860673-29860673 on Build GRCh38
rs781489225 Chr.16: 29860651-29860651 on Build GRCh38
rs778955484 Chr.16: 29860543-29860543 on Build GRCh38
rs1010341419 Chr.16: 29860528-29860528 on Build GRCh38
rs762425265 Chr.16: 29860692-29860692 on Build GRCh38
rs1041207819 Chr.16: 29860631-29860631 on Build GRCh38
rs368355579 Chr.16: 29860703-29860703 on Build GRCh38
rs746133972 Chr.16: 29860546-29860546 on Build GRCh38
rs772790263 Chr.16: 29860652-29860652 on Build GRCh38
rs1035053208 Chr.16: 29860625-29860625 on Build GRCh38
rs781055169 Chr.16: 29860632-29860632 on Build GRCh38
rs779675840 Chr.16: 29860582-29860582 on Build GRCh38
rs769312392 Chr.16: 29860590-29860590 on Build GRCh38
rs375184092 Chr.16: 29860601-29860601 on Build GRCh38
rs1050109566 Chr.16: 29860666-29860666 on Build GRCh38
rs199927687 Chr.16: 29860620-29860620 on Build GRCh38
rs372533371 Chr.16: 29860532-29860532 on Build GRCh38
rs771699178 Chr.16: 29860551-29860551 on Build GRCh38
rs192943434 Chr.16: 29860668-29860668 on Build GRCh38
rs776091165 Chr.16: 29860584-29860584 on Build GRCh38
rs201573714 Chr.16: 29860626-29860626 on Build GRCh38
rs565309751 Chr.16: 29860578-29860578 on Build GRCh38
rs762669095 Chr.16: 29860596-29860596 on Build GRCh38
rs758706964 Chr.16: 29860617-29860617 on Build GRCh38
rs762498266 Chr.16: 29860653-29860653 on Build GRCh38
rs761611134 Chr.16: 29860632-29860633 on Build GRCh38
rs775685296 Chr.16: 29860655-29860655 on Build GRCh38
rs779913485 Chr.16: 29860565-29860565 on Build GRCh38
rs748111316 Chr.16: 29860645-29860645 on Build GRCh38
rs772697464 Chr.16: 29860591-29860591 on Build GRCh38
rs114923885 Chr.16: 29860585-29860585 on Build GRCh38
rs113404632 Chr.16: 29860555-29860555 on Build GRCh38
rs962985880 Chr.16: 29860639-29860639 on Build GRCh38
rs768650553 Chr.16: 29860583-29860583 on Build GRCh38
rs753162397 Chr.16: 29860600-29860600 on Build GRCh38
rs1048902011 Chr.16: 29860609-29860609 on Build GRCh38

View Full Product Details