Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12641863-12642124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBL2
Entrez Gene ID
1389
Gene Name
cAMP responsive element binding protein like 2
Gene Aliases
-
UniGene
Hs.591156
Gene Chromosome Location
Chr.12: 12611833-12645108 on Build GRCh38
Transcripts
NM_001310.3

SNP Detail

SNP ID Chromosome Location
rs972852232 Chr.12: 12641898-12641898 on Build GRCh38
rs370560346 Chr.12: 12641947-12641947 on Build GRCh38
rs752051971 Chr.12: 12641905-12641905 on Build GRCh38
rs75440439 Chr.12: 12642053-12642053 on Build GRCh38
rs769020356 Chr.12: 12641982-12641982 on Build GRCh38
rs769813642 Chr.12: 12641948-12641948 on Build GRCh38
rs758113879 Chr.12: 12642038-12642038 on Build GRCh38
rs964897529 Chr.12: 12641964-12641964 on Build GRCh38
rs773720107 Chr.12: 12642016-12642016 on Build GRCh38
rs374724596 Chr.12: 12642047-12642047 on Build GRCh38
rs759148992 Chr.12: 12642025-12642025 on Build GRCh38
rs1010903282 Chr.12: 12642064-12642064 on Build GRCh38
rs939490530 Chr.12: 12641912-12641912 on Build GRCh38
rs141531026 Chr.12: 12642033-12642033 on Build GRCh38
rs764775373 Chr.12: 12642026-12642026 on Build GRCh38
rs774564799 Chr.12: 12641983-12641983 on Build GRCh38
rs200274326 Chr.12: 12641971-12641971 on Build GRCh38
rs767888899 Chr.12: 12642004-12642004 on Build GRCh38
rs928166557 Chr.12: 12641906-12641906 on Build GRCh38
rs181307517 Chr.12: 12642057-12642057 on Build GRCh38
rs545700004 Chr.12: 12641916-12641916 on Build GRCh38
rs1042873332 Chr.12: 12642093-12642093 on Build GRCh38
rs188490884 Chr.12: 12641923-12641923 on Build GRCh38
rs923557125 Chr.12: 12641999-12641999 on Build GRCh38
rs563854700 Chr.12: 12641917-12641917 on Build GRCh38
rs1009129250 Chr.12: 12642034-12642034 on Build GRCh38

View Full Product Details