Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110193097-110193358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
UniGene
Hs.428147
Gene Chromosome Location
Chr.6: 110180421-110232220 on Build GRCh38
Transcripts
NM_015891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3619446 c.211C>T p.H71Y Chr.6: 110193203-110193203 on Build GRCh38
6624245 c.254A>G p.Y85C Chr.6: 110193246-110193246 on Build GRCh38
4670675 c.225C>T p.A75A Chr.6: 110193217-110193217 on Build GRCh38
6624249 c.190-7delT p.? Chr.6: 110193175-110193175 on Build GRCh38
6624246 c.254A>G p.Y85C Chr.6: 110193246-110193246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748325407 Chr.6: 110193284-110193284 on Build GRCh38
rs1042439579 Chr.6: 110193220-110193220 on Build GRCh38
rs188381453 Chr.6: 110193217-110193217 on Build GRCh38
rs935478990 Chr.6: 110193221-110193221 on Build GRCh38
rs758034120 Chr.6: 110193161-110193161 on Build GRCh38
rs756904371 Chr.6: 110193218-110193218 on Build GRCh38
rs763702009 Chr.6: 110193310-110193310 on Build GRCh38
rs777388443 Chr.6: 110193309-110193310 on Build GRCh38
rs759791971 Chr.6: 110193296-110193296 on Build GRCh38
rs556916492 Chr.6: 110193239-110193239 on Build GRCh38
rs1049533900 Chr.6: 110193253-110193253 on Build GRCh38
rs761187443 Chr.6: 110193307-110193307 on Build GRCh38
rs936387655 Chr.6: 110193240-110193240 on Build GRCh38
rs202176812 Chr.6: 110193203-110193203 on Build GRCh38
rs760114740 Chr.6: 110193137-110193137 on Build GRCh38
rs774661776 Chr.6: 110193132-110193132 on Build GRCh38
rs755576410 Chr.6: 110193165-110193165 on Build GRCh38
rs778881212 Chr.6: 110193238-110193238 on Build GRCh38
rs779821764 Chr.6: 110193246-110193246 on Build GRCh38
rs769745679 Chr.6: 110193251-110193251 on Build GRCh38
rs201325988 Chr.6: 110193279-110193279 on Build GRCh38
rs377292914 Chr.6: 110193161-110193161 on Build GRCh38
rs768137860 Chr.6: 110193302-110193302 on Build GRCh38
rs367731613 Chr.6: 110193306-110193306 on Build GRCh38
rs773215022 Chr.6: 110193254-110193254 on Build GRCh38
rs766112969 Chr.6: 110193158-110193158 on Build GRCh38
rs889260518 Chr.6: 110193296-110193296 on Build GRCh38
rs758456603 Chr.6: 110193154-110193154 on Build GRCh38
rs774749469 Chr.6: 110193283-110193283 on Build GRCh38
rs749546724 Chr.6: 110193257-110193257 on Build GRCh38
rs371145621 Chr.6: 110193214-110193214 on Build GRCh38
rs576621877 Chr.6: 110193247-110193247 on Build GRCh38

View Full Product Details