Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10556942-10557211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT2
Entrez Gene ID
2651
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
UniGene
Hs.519884
Gene Chromosome Location
Chr.6: 10521283-10629368 on Build GRCh38
Transcripts
NM_001491.2 NM_145649.4 XM_005248997.3 XM_005248999.2 XM_006715052.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5733935 c.754_755insC p.H254fs*2 Chr.6: 10557177-10557178 on Build GRCh38
5404096 c.785C>T p.S262F Chr.6: 10557208-10557208 on Build GRCh38
1439497 c.754delC p.H254fs*47 Chr.6: 10557177-10557177 on Build GRCh38
5522748 c.523C>T p.L175F Chr.6: 10556946-10556946 on Build GRCh38
1642818 c.586delA p.T197fs*5 Chr.6: 10557009-10557009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769830102 Chr.6: 10557152-10557154 on Build GRCh38
rs200427106 Chr.6: 10557145-10557145 on Build GRCh38
rs756141442 Chr.6: 10557014-10557014 on Build GRCh38
rs752425717 Chr.6: 10556977-10556977 on Build GRCh38
rs763988372 Chr.6: 10556981-10556981 on Build GRCh38
rs932074587 Chr.6: 10557012-10557012 on Build GRCh38
rs911875612 Chr.6: 10557028-10557028 on Build GRCh38
rs374053208 Chr.6: 10557182-10557182 on Build GRCh38
rs201670024 Chr.6: 10557002-10557002 on Build GRCh38
rs773541420 Chr.6: 10557136-10557136 on Build GRCh38
rs758926587 Chr.6: 10557072-10557072 on Build GRCh38
rs775131144 Chr.6: 10557155-10557155 on Build GRCh38
rs756999080 Chr.6: 10557176-10557177 on Build GRCh38
rs557486400 Chr.6: 10557007-10557007 on Build GRCh38
rs748603355 Chr.6: 10557048-10557048 on Build GRCh38
rs868361696 Chr.6: 10557135-10557135 on Build GRCh38
rs865926878 Chr.6: 10557000-10557001 on Build GRCh38
rs757132008 Chr.6: 10556995-10556995 on Build GRCh38
rs200874437 Chr.6: 10557164-10557164 on Build GRCh38
rs143057896 Chr.6: 10557132-10557132 on Build GRCh38
rs751471040 Chr.6: 10556988-10556988 on Build GRCh38
rs553300905 Chr.6: 10557073-10557073 on Build GRCh38
rs766359774 Chr.6: 10557181-10557181 on Build GRCh38
rs756250960 Chr.6: 10557005-10557005 on Build GRCh38
rs779151758 Chr.6: 10557117-10557117 on Build GRCh38
rs150240128 Chr.6: 10557013-10557013 on Build GRCh38
rs949335475 Chr.6: 10557133-10557134 on Build GRCh38
rs572831032 Chr.6: 10557025-10557025 on Build GRCh38
rs764763524 Chr.6: 10556976-10556976 on Build GRCh38
rs745904861 Chr.6: 10557003-10557003 on Build GRCh38
rs748426560 Chr.6: 10557118-10557118 on Build GRCh38
rs574686656 Chr.6: 10557087-10557087 on Build GRCh38
rs944967206 Chr.6: 10557049-10557049 on Build GRCh38
rs754406768 Chr.6: 10557168-10557168 on Build GRCh38
rs148372193 Chr.6: 10557053-10557053 on Build GRCh38
rs757116901 Chr.6: 10557099-10557099 on Build GRCh38
rs756088362 Chr.6: 10557106-10557106 on Build GRCh38
rs768135788 Chr.6: 10556972-10556972 on Build GRCh38
rs775219051 Chr.6: 10557080-10557080 on Build GRCh38
rs931610524 Chr.6: 10557033-10557033 on Build GRCh38
rs749436278 Chr.6: 10557111-10557111 on Build GRCh38
rs769030276 Chr.6: 10556963-10556963 on Build GRCh38
rs921622046 Chr.6: 10557030-10557030 on Build GRCh38
rs752982694 Chr.6: 10557183-10557183 on Build GRCh38
rs369284218 Chr.6: 10557081-10557081 on Build GRCh38
rs199596076 Chr.6: 10557163-10557163 on Build GRCh38
rs762604882 Chr.6: 10557156-10557156 on Build GRCh38
rs758212318 Chr.6: 10556978-10556978 on Build GRCh38
rs564300384 Chr.6: 10557178-10557178 on Build GRCh38
rs199770755 Chr.6: 10556975-10556975 on Build GRCh38
rs751290591 Chr.6: 10557095-10557095 on Build GRCh38
rs35318346 Chr.6: 10557110-10557110 on Build GRCh38
rs774780221 Chr.6: 10556967-10556967 on Build GRCh38
rs755180639 Chr.6: 10557112-10557112 on Build GRCh38
rs764139154 Chr.6: 10557100-10557100 on Build GRCh38
rs762230655 Chr.6: 10556971-10556971 on Build GRCh38
rs370701306 Chr.6: 10557173-10557173 on Build GRCh38
rs774692478 Chr.6: 10557042-10557042 on Build GRCh38
rs372757154 Chr.6: 10557149-10557149 on Build GRCh38
rs763598223 Chr.6: 10557001-10557002 on Build GRCh38
rs773805726 Chr.6: 10557060-10557060 on Build GRCh38

View Full Product Details