Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33583377-33583618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6648602 c.934C>T p.R312* Chr.21: 33583518-33583518 on Build GRCh38
4752623 c.917T>A p.L306H Chr.21: 33583535-33583535 on Build GRCh38
3363430 c.838T>C p.Y280H Chr.21: 33583614-33583614 on Build GRCh38
7250812 c.896G>T p.G299V Chr.21: 33583556-33583556 on Build GRCh38
4399918 c.909C>T p.I303I Chr.21: 33583543-33583543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542296982 Chr.21: 33583575-33583575 on Build GRCh38
rs776562020 Chr.21: 33583587-33583587 on Build GRCh38
rs757469136 Chr.21: 33583556-33583556 on Build GRCh38
rs933956443 Chr.21: 33583432-33583432 on Build GRCh38
rs778867526 Chr.21: 33583455-33583455 on Build GRCh38
rs765047518 Chr.21: 33583513-33583513 on Build GRCh38
rs774841815 Chr.21: 33583496-33583497 on Build GRCh38
rs753995684 Chr.21: 33583537-33583537 on Build GRCh38
rs929871493 Chr.21: 33583409-33583409 on Build GRCh38
rs768592814 Chr.21: 33583476-33583476 on Build GRCh38
rs146301000 Chr.21: 33583533-33583533 on Build GRCh38
rs899854707 Chr.21: 33583426-33583426 on Build GRCh38
rs762575148 Chr.21: 33583531-33583531 on Build GRCh38
rs1017127374 Chr.21: 33583492-33583492 on Build GRCh38
rs534744181 Chr.21: 33583439-33583439 on Build GRCh38
rs759427178 Chr.21: 33583577-33583577 on Build GRCh38
rs769216480 Chr.21: 33583477-33583478 on Build GRCh38
rs779803447 Chr.21: 33583576-33583576 on Build GRCh38
rs771879018 Chr.21: 33583461-33583461 on Build GRCh38
rs898637106 Chr.21: 33583493-33583493 on Build GRCh38
rs757636594 Chr.21: 33583574-33583574 on Build GRCh38
rs770942339 Chr.21: 33583452-33583452 on Build GRCh38
rs768499054 Chr.21: 33583583-33583583 on Build GRCh38
rs187959367 Chr.21: 33583475-33583475 on Build GRCh38
rs182381618 Chr.21: 33583442-33583442 on Build GRCh38
rs761560613 Chr.21: 33583503-33583503 on Build GRCh38
rs775533543 Chr.21: 33583464-33583464 on Build GRCh38
rs148095112 Chr.21: 33583560-33583560 on Build GRCh38
rs756425637 Chr.21: 33583536-33583536 on Build GRCh38
rs776365674 Chr.21: 33583478-33583478 on Build GRCh38
rs905943086 Chr.21: 33583540-33583540 on Build GRCh38
rs751927127 Chr.21: 33583517-33583517 on Build GRCh38
rs746015074 Chr.21: 33583457-33583457 on Build GRCh38

View Full Product Details