Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50314000-50314271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND6B
Entrez Gene ID
414918
Gene Name
DENN domain containing 6B
Gene Aliases
AFI1B, FAM116B
UniGene
Hs.530380
Gene Chromosome Location
Chr.22: 50309032-50327088 on Build GRCh38
Transcripts
NM_001001794.3 XM_005261914.3 XM_011530692.2 XM_011530693.2 XM_017028801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5447489 c.1087C>T p.Q363* Chr.22: 50314258-50314258 on Build GRCh38
4827664 c.1119G>A p.K373K Chr.22: 50314226-50314226 on Build GRCh38
5016694 c.1086_1101del16 p.Q363fs*5 Chr.22: 50314244-50314259 on Build GRCh38
3371956 c.1116G>A p.L372L Chr.22: 50314229-50314229 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374419955 Chr.22: 50314172-50314172 on Build GRCh38
rs551514259 Chr.22: 50314109-50314109 on Build GRCh38
rs772756513 Chr.22: 50314224-50314224 on Build GRCh38
rs778613236 Chr.22: 50314137-50314137 on Build GRCh38
rs562198369 Chr.22: 50314044-50314044 on Build GRCh38
rs552059657 Chr.22: 50314135-50314135 on Build GRCh38
rs1055066532 Chr.22: 50314223-50314223 on Build GRCh38
rs911086586 Chr.22: 50314083-50314083 on Build GRCh38
rs986407271 Chr.22: 50314155-50314155 on Build GRCh38
rs757546034 Chr.22: 50314181-50314181 on Build GRCh38
rs761278399 Chr.22: 50314177-50314177 on Build GRCh38
rs111779373 Chr.22: 50314147-50314147 on Build GRCh38
rs369498737 Chr.22: 50314233-50314233 on Build GRCh38
rs888444628 Chr.22: 50314136-50314136 on Build GRCh38
rs760474291 Chr.22: 50314238-50314238 on Build GRCh38
rs1042479537 Chr.22: 50314235-50314235 on Build GRCh38
rs758339557 Chr.22: 50314192-50314192 on Build GRCh38
rs368989461 Chr.22: 50314160-50314160 on Build GRCh38
rs552925286 Chr.22: 50314154-50314154 on Build GRCh38
rs374286615 Chr.22: 50314119-50314119 on Build GRCh38
rs1049925971 Chr.22: 50314076-50314076 on Build GRCh38
rs546918694 Chr.22: 50314038-50314038 on Build GRCh38
rs988438771 Chr.22: 50314046-50314046 on Build GRCh38
rs1037808785 Chr.22: 50314021-50314021 on Build GRCh38
rs910514619 Chr.22: 50314156-50314156 on Build GRCh38
rs765433530 Chr.22: 50314191-50314191 on Build GRCh38
rs771691447 Chr.22: 50314179-50314179 on Build GRCh38
rs560021955 Chr.22: 50314146-50314146 on Build GRCh38
rs927792385 Chr.22: 50314189-50314189 on Build GRCh38
rs919361183 Chr.22: 50314033-50314033 on Build GRCh38
rs970805819 Chr.22: 50314170-50314170 on Build GRCh38
rs893487666 Chr.22: 50314115-50314115 on Build GRCh38
rs982411863 Chr.22: 50314022-50314022 on Build GRCh38
rs752558999 Chr.22: 50314210-50314210 on Build GRCh38
rs749142263 Chr.22: 50314247-50314247 on Build GRCh38
rs746372899 Chr.22: 50314034-50314034 on Build GRCh38
rs749841275 Chr.22: 50314125-50314125 on Build GRCh38
rs536909297 Chr.22: 50314132-50314132 on Build GRCh38
rs756089259 Chr.22: 50314221-50314221 on Build GRCh38
rs1055167304 Chr.22: 50314104-50314104 on Build GRCh38
rs775697398 Chr.22: 50314129-50314130 on Build GRCh38
rs754293774 Chr.22: 50314180-50314180 on Build GRCh38
rs764973258 Chr.22: 50314241-50314241 on Build GRCh38
rs190906774 Chr.22: 50314173-50314173 on Build GRCh38
rs758919092 Chr.22: 50314166-50314166 on Build GRCh38
rs780210515 Chr.22: 50314208-50314208 on Build GRCh38
rs550534717 Chr.22: 50314107-50314107 on Build GRCh38
rs1002126045 Chr.22: 50314058-50314058 on Build GRCh38
rs529397900 Chr.22: 50314057-50314057 on Build GRCh38
rs752014240 Chr.22: 50314179-50314179 on Build GRCh38
rs914203585 Chr.22: 50314054-50314054 on Build GRCh38

View Full Product Details