Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68934324-68934563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX50
Entrez Gene ID
79009
Gene Name
DEAD-box helicase 50
Gene Aliases
GU2, GUB, RH-II/GuB, mcdrh
UniGene
Hs.522984
Gene Chromosome Location
Chr.10: 68901278-68956312 on Build GRCh38
Transcripts
NM_024045.1 XM_005270148.2 XM_011540144.2 XM_017016626.1 XM_017016627.1 XM_017016628.1 XM_017016629.1 XM_017016630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6847458 c.1396C>A p.P466T Chr.10: 68934355-68934355 on Build GRCh38
6288458 c.1387A>G p.S463G Chr.10: 68934346-68934346 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372411914 Chr.10: 68934357-68934357 on Build GRCh38
rs954692964 Chr.10: 68934506-68934506 on Build GRCh38
rs952206280 Chr.10: 68934389-68934389 on Build GRCh38
rs748336693 Chr.10: 68934380-68934380 on Build GRCh38
rs756260784 Chr.10: 68934349-68934349 on Build GRCh38
rs974625816 Chr.10: 68934475-68934476 on Build GRCh38
rs775104191 Chr.10: 68934387-68934387 on Build GRCh38
rs779054661 Chr.10: 68934351-68934351 on Build GRCh38
rs1021854077 Chr.10: 68934413-68934413 on Build GRCh38
rs968583744 Chr.10: 68934425-68934425 on Build GRCh38
rs771183158 Chr.10: 68934435-68934435 on Build GRCh38
rs907846823 Chr.10: 68934534-68934534 on Build GRCh38
rs987494651 Chr.10: 68934516-68934516 on Build GRCh38
rs940733461 Chr.10: 68934535-68934535 on Build GRCh38
rs114610405 Chr.10: 68934409-68934409 on Build GRCh38
rs746692243 Chr.10: 68934405-68934405 on Build GRCh38
rs1028950870 Chr.10: 68934498-68934498 on Build GRCh38
rs771901959 Chr.10: 68934362-68934362 on Build GRCh38
rs544443927 Chr.10: 68934466-68934466 on Build GRCh38
rs547005058 Chr.10: 68934430-68934430 on Build GRCh38

View Full Product Details