Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168308632-168308887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT2
Entrez Gene ID
168002
Gene Name
dishevelled binding antagonist of beta catenin 2
Gene Aliases
C6orf116, DAPPER2, DPR2, bA503C24.7
UniGene
Hs.673059
Gene Chromosome Location
Chr.6: 168292830-168319754 on Build GRCh38
Transcripts
NM_001286350.1 NM_001286351.1 NM_214462.4 NR_104425.1 XM_011535507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4635362 c.1116T>C p.S372S Chr.6: 168308641-168308641 on Build GRCh38
1487473 c.1051G>A p.E351K Chr.6: 168308706-168308706 on Build GRCh38
5369208 c.895G>A p.G299S Chr.6: 168308862-168308862 on Build GRCh38
5447890 c.1091C>T p.S364F Chr.6: 168308666-168308666 on Build GRCh38
5737845 c.980C>T p.P327L Chr.6: 168308777-168308777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760355743 Chr.6: 168308730-168308730 on Build GRCh38
rs868612517 Chr.6: 168308796-168308796 on Build GRCh38
rs781216476 Chr.6: 168308686-168308686 on Build GRCh38
rs895766632 Chr.6: 168308809-168308809 on Build GRCh38
rs35627974 Chr.6: 168308848-168308848 on Build GRCh38
rs570102629 Chr.6: 168308795-168308795 on Build GRCh38
rs777886087 Chr.6: 168308790-168308790 on Build GRCh38
rs766517269 Chr.6: 168308722-168308722 on Build GRCh38
rs918776019 Chr.6: 168308757-168308757 on Build GRCh38
rs747216419 Chr.6: 168308794-168308794 on Build GRCh38
rs754693657 Chr.6: 168308851-168308851 on Build GRCh38
rs911540065 Chr.6: 168308750-168308750 on Build GRCh38
rs775375348 Chr.6: 168308684-168308684 on Build GRCh38
rs559934089 Chr.6: 168308774-168308774 on Build GRCh38
rs867445231 Chr.6: 168308678-168308678 on Build GRCh38
rs532586396 Chr.6: 168308755-168308755 on Build GRCh38
rs867440507 Chr.6: 168308762-168308762 on Build GRCh38
rs757463036 Chr.6: 168308674-168308674 on Build GRCh38
rs769524155 Chr.6: 168308675-168308675 on Build GRCh38
rs531256391 Chr.6: 168308842-168308842 on Build GRCh38
rs953090262 Chr.6: 168308725-168308725 on Build GRCh38
rs367909442 Chr.6: 168308651-168308651 on Build GRCh38
rs541416019 Chr.6: 168308756-168308756 on Build GRCh38
rs1000054867 Chr.6: 168308852-168308852 on Build GRCh38
rs1042000197 Chr.6: 168308827-168308827 on Build GRCh38
rs374278981 Chr.6: 168308705-168308705 on Build GRCh38
rs763015294 Chr.6: 168308696-168308696 on Build GRCh38
rs766679635 Chr.6: 168308754-168308754 on Build GRCh38
rs905245186 Chr.6: 168308806-168308806 on Build GRCh38
rs1028828205 Chr.6: 168308861-168308861 on Build GRCh38
rs935216597 Chr.6: 168308763-168308763 on Build GRCh38
rs1045582968 Chr.6: 168308802-168308802 on Build GRCh38
rs753191558 Chr.6: 168308787-168308787 on Build GRCh38
rs767045985 Chr.6: 168308713-168308713 on Build GRCh38
rs757460539 Chr.6: 168308805-168308805 on Build GRCh38
rs530127614 Chr.6: 168308788-168308788 on Build GRCh38
rs937417037 Chr.6: 168308800-168308800 on Build GRCh38
rs201471118 Chr.6: 168308824-168308824 on Build GRCh38
rs939366648 Chr.6: 168308823-168308823 on Build GRCh38
rs6925614 Chr.6: 168308705-168308705 on Build GRCh38
rs765675691 Chr.6: 168308776-168308776 on Build GRCh38
rs548772905 Chr.6: 168308792-168308792 on Build GRCh38
rs565295919 Chr.6: 168308729-168308729 on Build GRCh38
rs774006853 Chr.6: 168308706-168308706 on Build GRCh38
rs1020120816 Chr.6: 168308664-168308664 on Build GRCh38

View Full Product Details