Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210587981-210588222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
UniGene
Hs.149252
Gene Chromosome Location
Chr.2: 210477682-210679107 on Build GRCh38
Transcripts
NM_001122633.2 NM_001875.4 XM_011510640.1 XM_011510641.2 XM_011510642.2 XM_011510643.2 XM_011510644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7347478 c.676G>C p.V226L Chr.2: 210588094-210588094 on Build GRCh38
371847 c.627C>A p.V209V Chr.2: 210588063-210588063 on Build GRCh38
5500597 c.692T>C p.V231A Chr.2: 210588128-210588128 on Build GRCh38
3577067 c.661G>A p.G221R Chr.2: 210588079-210588079 on Build GRCh38
3838482 c.622-1G>A p.? Chr.2: 210588057-210588057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746845709 Chr.2: 210588043-210588043 on Build GRCh38
rs767905306 Chr.2: 210588133-210588133 on Build GRCh38
rs766500766 Chr.2: 210588176-210588176 on Build GRCh38
rs370984582 Chr.2: 210588184-210588184 on Build GRCh38
rs762550873 Chr.2: 210588094-210588094 on Build GRCh38
rs756703955 Chr.2: 210588154-210588154 on Build GRCh38
rs745365017 Chr.2: 210588063-210588063 on Build GRCh38
rs374275578 Chr.2: 210588132-210588132 on Build GRCh38
rs369210668 Chr.2: 210588021-210588021 on Build GRCh38
rs376312096 Chr.2: 210588178-210588178 on Build GRCh38
rs374922944 Chr.2: 210588180-210588180 on Build GRCh38
rs74775355 Chr.2: 210588053-210588053 on Build GRCh38
rs1053621627 Chr.2: 210588167-210588167 on Build GRCh38
rs774556324 Chr.2: 210588103-210588103 on Build GRCh38
rs762276786 Chr.2: 210588126-210588126 on Build GRCh38
rs752090554 Chr.2: 210588176-210588176 on Build GRCh38
rs763828318 Chr.2: 210588095-210588095 on Build GRCh38
rs894978816 Chr.2: 210588182-210588182 on Build GRCh38
rs138724401 Chr.2: 210588072-210588072 on Build GRCh38
rs757147536 Chr.2: 210588191-210588191 on Build GRCh38
rs1000750975 Chr.2: 210588147-210588147 on Build GRCh38
rs776595181 Chr.2: 210588060-210588060 on Build GRCh38
rs777393786 Chr.2: 210588029-210588029 on Build GRCh38
rs112357214 Chr.2: 210588054-210588054 on Build GRCh38
rs769324029 Chr.2: 210588091-210588091 on Build GRCh38
rs375610226 Chr.2: 210588185-210588185 on Build GRCh38
rs751029486 Chr.2: 210588134-210588134 on Build GRCh38
rs1049458960 Chr.2: 210588027-210588027 on Build GRCh38
rs138392504 Chr.2: 210588073-210588073 on Build GRCh38

View Full Product Details