Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154837670-154837895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
UniGene
Hs.632836
Gene Chromosome Location
Chr.X: 154835788-155022723 on Build GRCh38
Transcripts
NM_000132.3 NM_019863.2

SNP Detail

SNP ID Chromosome Location
rs373079141 Chr.X: 154837724-154837724 on Build GRCh38
rs782799573 Chr.X: 154837702-154837702 on Build GRCh38
rs137852472 Chr.X: 154837697-154837697 on Build GRCh38
rs782741405 Chr.X: 154837841-154837841 on Build GRCh38
rs782489626 Chr.X: 154837705-154837705 on Build GRCh38
rs1005867077 Chr.X: 154837852-154837852 on Build GRCh38
rs782621263 Chr.X: 154837711-154837711 on Build GRCh38
rs137852374 Chr.X: 154837698-154837698 on Build GRCh38
rs782053499 Chr.X: 154837801-154837801 on Build GRCh38
rs938562671 Chr.X: 154837695-154837695 on Build GRCh38
rs192393197 Chr.X: 154837780-154837780 on Build GRCh38
rs1047644991 Chr.X: 154837721-154837721 on Build GRCh38
rs387906466 Chr.X: 154837737-154837741 on Build GRCh38
rs886359315 Chr.X: 154837752-154837752 on Build GRCh38
rs782459456 Chr.X: 154837760-154837760 on Build GRCh38
rs200902529 Chr.X: 154837696-154837696 on Build GRCh38

View Full Product Details