Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130563532-130563787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A6
Entrez Gene ID
131873
Gene Name
collagen type VI alpha 6
Gene Aliases
-
UniGene
Hs.591282
Gene Chromosome Location
Chr.3: 130516709-130678135 on Build GRCh38
Transcripts
NM_001102608.1 XM_005247121.4 XM_011512423.2 XM_011512424.2 XM_011512425.2 XM_011512426.2 XM_011512428.2 XM_011512431.2 XM_011512432.2 XM_017005711.1 XM_017005712.1 XM_017005713.1 XM_017005714.1 XM_017005715.1 XM_017005716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544817 c.552C>T p.F184F Chr.3: 130563555-130563555 on Build GRCh38
5398156 c.613G>A p.D205N Chr.3: 130563616-130563616 on Build GRCh38
5708467 c.568A>G p.R190G Chr.3: 130563571-130563571 on Build GRCh38
3587345 c.547C>T p.H183Y Chr.3: 130563550-130563550 on Build GRCh38
3427057 c.651C>A p.I217I Chr.3: 130563654-130563654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978855766 Chr.3: 130563604-130563604 on Build GRCh38
rs1027132069 Chr.3: 130563612-130563612 on Build GRCh38
rs766676671 Chr.3: 130563608-130563608 on Build GRCh38
rs375368589 Chr.3: 130563660-130563660 on Build GRCh38
rs1006324562 Chr.3: 130563669-130563669 on Build GRCh38
rs768036587 Chr.3: 130563601-130563601 on Build GRCh38
rs141494444 Chr.3: 130563657-130563657 on Build GRCh38
rs1014626451 Chr.3: 130563640-130563640 on Build GRCh38
rs200264014 Chr.3: 130563562-130563562 on Build GRCh38
rs1046405934 Chr.3: 130563759-130563759 on Build GRCh38
rs1035611775 Chr.3: 130563728-130563728 on Build GRCh38
rs771428493 Chr.3: 130563709-130563709 on Build GRCh38
rs375675387 Chr.3: 130563703-130563703 on Build GRCh38
rs915677885 Chr.3: 130563599-130563599 on Build GRCh38
rs376108254 Chr.3: 130563607-130563607 on Build GRCh38
rs993337843 Chr.3: 130563727-130563727 on Build GRCh38
rs746883936 Chr.3: 130563653-130563653 on Build GRCh38
rs959856044 Chr.3: 130563760-130563760 on Build GRCh38
rs867631537 Chr.3: 130563751-130563751 on Build GRCh38
rs765348963 Chr.3: 130563632-130563632 on Build GRCh38
rs758510881 Chr.3: 130563643-130563643 on Build GRCh38
rs949353027 Chr.3: 130563742-130563742 on Build GRCh38
rs150427289 Chr.3: 130563666-130563666 on Build GRCh38
rs79600490 Chr.3: 130563704-130563704 on Build GRCh38
rs373400548 Chr.3: 130563624-130563624 on Build GRCh38
rs752841549 Chr.3: 130563634-130563634 on Build GRCh38
rs561667088 Chr.3: 130563724-130563724 on Build GRCh38
rs779524794 Chr.3: 130563692-130563692 on Build GRCh38
rs78652791 Chr.3: 130563723-130563723 on Build GRCh38
rs372218106 Chr.3: 130563702-130563702 on Build GRCh38
rs775103973 Chr.3: 130563577-130563577 on Build GRCh38
rs190071598 Chr.3: 130563659-130563659 on Build GRCh38
rs762349200 Chr.3: 130563584-130563584 on Build GRCh38
rs763117396 Chr.3: 130563567-130563567 on Build GRCh38
rs371520731 Chr.3: 130563623-130563623 on Build GRCh38
rs185319822 Chr.3: 130563563-130563563 on Build GRCh38
rs773669480 Chr.3: 130563605-130563605 on Build GRCh38
rs754144093 Chr.3: 130563617-130563617 on Build GRCh38
rs543636468 Chr.3: 130563568-130563568 on Build GRCh38
rs745592388 Chr.3: 130563570-130563570 on Build GRCh38
rs953103750 Chr.3: 130563674-130563674 on Build GRCh38
rs769535987 Chr.3: 130563671-130563671 on Build GRCh38
rs377089245 Chr.3: 130563644-130563644 on Build GRCh38
rs769325649 Chr.3: 130563572-130563572 on Build GRCh38
rs368044533 Chr.3: 130563693-130563693 on Build GRCh38
rs531799741 Chr.3: 130563729-130563729 on Build GRCh38

View Full Product Details