Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152829074-152829347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CETN2
Entrez Gene ID
1069
Gene Name
centrin 2
Gene Aliases
CALT, CEN2
UniGene
Hs.82794
Gene Chromosome Location
Chr.X: 152827327-152830757 on Build GRCh38
Transcripts
NM_004344.1

SNP Detail

SNP ID Chromosome Location
rs782593030 Chr.X: 152829125-152829125 on Build GRCh38
rs782340354 Chr.X: 152829145-152829145 on Build GRCh38
rs782285029 Chr.X: 152829135-152829135 on Build GRCh38
rs375684661 Chr.X: 152829317-152829317 on Build GRCh38
rs782438296 Chr.X: 152829138-152829138 on Build GRCh38
rs782260978 Chr.X: 152829141-152829141 on Build GRCh38
rs374235428 Chr.X: 152829180-152829180 on Build GRCh38
rs781978644 Chr.X: 152829118-152829118 on Build GRCh38
rs141784522 Chr.X: 152829284-152829284 on Build GRCh38
rs781816418 Chr.X: 152829271-152829271 on Build GRCh38
rs782106968 Chr.X: 152829203-152829203 on Build GRCh38
rs927332703 Chr.X: 152829195-152829195 on Build GRCh38
rs370304608 Chr.X: 152829140-152829140 on Build GRCh38
rs368139654 Chr.X: 152829197-152829197 on Build GRCh38
rs782068050 Chr.X: 152829200-152829200 on Build GRCh38
rs782787803 Chr.X: 152829231-152829231 on Build GRCh38
rs782543051 Chr.X: 152829319-152829319 on Build GRCh38
rs11542846 Chr.X: 152829260-152829260 on Build GRCh38
rs781795967 Chr.X: 152829134-152829134 on Build GRCh38
rs782049461 Chr.X: 152829139-152829139 on Build GRCh38
rs782689752 Chr.X: 152829320-152829320 on Build GRCh38
rs782033927 Chr.X: 152829201-152829201 on Build GRCh38

View Full Product Details