Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74716510-74716781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FA2H
Entrez Gene ID
79152
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
FAAH, FAH1, FAXDC1, SCS7, SPG35
UniGene
Hs.461329
Gene Chromosome Location
Chr.16: 74712955-74774831 on Build GRCh38
Transcripts
NM_024306.4 XM_011523317.2 XM_011523319.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4414327 c.816C>G p.F272L Chr.16: 74716570-74716570 on Build GRCh38
6032918 c.861C>T p.C287C Chr.16: 74716525-74716525 on Build GRCh38
5530747 c.852C>T p.F284F Chr.16: 74716534-74716534 on Build GRCh38
973662 c.795C>T p.F265F Chr.16: 74716591-74716591 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530195358 Chr.16: 74716613-74716613 on Build GRCh38
rs899587066 Chr.16: 74716603-74716603 on Build GRCh38
rs57844314 Chr.16: 74716658-74716658 on Build GRCh38
rs962932498 Chr.16: 74716657-74716657 on Build GRCh38
rs142644174 Chr.16: 74716583-74716583 on Build GRCh38
rs1024891051 Chr.16: 74716712-74716712 on Build GRCh38
rs747693437 Chr.16: 74716686-74716686 on Build GRCh38
rs779833110 Chr.16: 74716654-74716654 on Build GRCh38
rs890278057 Chr.16: 74716552-74716552 on Build GRCh38
rs944558398 Chr.16: 74716720-74716720 on Build GRCh38
rs1035158000 Chr.16: 74716621-74716621 on Build GRCh38
rs766896095 Chr.16: 74716708-74716708 on Build GRCh38
rs528121279 Chr.16: 74716688-74716688 on Build GRCh38
rs867086433 Chr.16: 74716568-74716568 on Build GRCh38
rs781362098 Chr.16: 74716530-74716530 on Build GRCh38
rs762169178 Chr.16: 74716595-74716595 on Build GRCh38
rs199815871 Chr.16: 74716542-74716542 on Build GRCh38
rs943671314 Chr.16: 74716581-74716581 on Build GRCh38
rs541507213 Chr.16: 74716555-74716555 on Build GRCh38
rs1052369680 Chr.16: 74716556-74716556 on Build GRCh38
rs762987810 Chr.16: 74716587-74716587 on Build GRCh38
rs757812676 Chr.16: 74716528-74716528 on Build GRCh38
rs906732910 Chr.16: 74716618-74716618 on Build GRCh38
rs868146622 Chr.16: 74716631-74716631 on Build GRCh38
rs544646155 Chr.16: 74716543-74716543 on Build GRCh38
rs1001933177 Chr.16: 74716604-74716604 on Build GRCh38
rs775735494 Chr.16: 74716572-74716572 on Build GRCh38
rs1052695357 Chr.16: 74716728-74716728 on Build GRCh38
rs892417434 Chr.16: 74716730-74716730 on Build GRCh38
rs886043274 Chr.16: 74716591-74716591 on Build GRCh38
rs112754063 Chr.16: 74716757-74716757 on Build GRCh38
rs921252006 Chr.16: 74716659-74716659 on Build GRCh38
rs771402018 Chr.16: 74716588-74716588 on Build GRCh38
rs149133141 Chr.16: 74716592-74716592 on Build GRCh38
rs370734744 Chr.16: 74716664-74716664 on Build GRCh38
rs778242149 Chr.16: 74716668-74716668 on Build GRCh38
rs1027603627 Chr.16: 74716732-74716732 on Build GRCh38
rs912602937 Chr.16: 74716713-74716713 on Build GRCh38
rs370973830 Chr.16: 74716540-74716540 on Build GRCh38
rs983322885 Chr.16: 74716645-74716645 on Build GRCh38
rs1017476556 Chr.16: 74716633-74716634 on Build GRCh38
rs373152103 Chr.16: 74716566-74716566 on Build GRCh38
rs374680337 Chr.16: 74716644-74716644 on Build GRCh38
rs143255996 Chr.16: 74716695-74716695 on Build GRCh38
rs750814402 Chr.16: 74716635-74716635 on Build GRCh38
rs773369721 Chr.16: 74716653-74716653 on Build GRCh38
rs138244546 Chr.16: 74716539-74716539 on Build GRCh38
rs754694789 Chr.16: 74716666-74716666 on Build GRCh38
rs572400760 Chr.16: 74716573-74716573 on Build GRCh38
rs374386471 Chr.16: 74716562-74716562 on Build GRCh38
rs1010615037 Chr.16: 74716584-74716584 on Build GRCh38
rs878855083 Chr.16: 74716564-74716565 on Build GRCh38
rs571370504 Chr.16: 74716748-74716748 on Build GRCh38

View Full Product Details