Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41094924-41095197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A13
Entrez Gene ID
1553
Gene Name
cytochrome P450 family 2 subfamily A member 13
Gene Aliases
CPAD, CYP2A, CYPIIA13
UniGene
Hs.567252
Gene Chromosome Location
Chr.19: 41088451-41096195 on Build GRCh38
Transcripts
NM_000766.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160304 c.1225C>T p.R409W Chr.19: 41095022-41095022 on Build GRCh38
6084929 c.1178C>T p.P393L Chr.19: 41094975-41094975 on Build GRCh38
3404260 c.1195C>T p.L399L Chr.19: 41094992-41094992 on Build GRCh38
1712328 c.1237C>T p.P413S Chr.19: 41095034-41095034 on Build GRCh38
6224906 c.1209G>A p.R403R Chr.19: 41095006-41095006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773564903 Chr.19: 41095000-41095000 on Build GRCh38
rs781121947 Chr.19: 41095031-41095031 on Build GRCh38
rs187762561 Chr.19: 41095088-41095088 on Build GRCh38
rs145584508 Chr.19: 41095052-41095052 on Build GRCh38
rs779688987 Chr.19: 41095099-41095099 on Build GRCh38
rs773217452 Chr.19: 41094975-41094975 on Build GRCh38
rs752229472 Chr.19: 41095073-41095073 on Build GRCh38
rs764280051 Chr.19: 41095176-41095176 on Build GRCh38
rs961481096 Chr.19: 41095175-41095175 on Build GRCh38
rs748853854 Chr.19: 41094960-41094960 on Build GRCh38
rs145039737 Chr.19: 41094988-41094988 on Build GRCh38
rs866002403 Chr.19: 41095017-41095017 on Build GRCh38
rs753400288 Chr.19: 41095069-41095069 on Build GRCh38
rs199846503 Chr.19: 41095026-41095026 on Build GRCh38
rs777504001 Chr.19: 41094958-41094958 on Build GRCh38
rs370801766 Chr.19: 41095015-41095015 on Build GRCh38
rs769819999 Chr.19: 41095136-41095136 on Build GRCh38
rs140338223 Chr.19: 41095054-41095054 on Build GRCh38
rs754869860 Chr.19: 41095130-41095130 on Build GRCh38
rs775232883 Chr.19: 41094970-41094970 on Build GRCh38
rs765002438 Chr.19: 41095078-41095078 on Build GRCh38
rs758249719 Chr.19: 41095091-41095091 on Build GRCh38
rs756963808 Chr.19: 41095019-41095019 on Build GRCh38
rs778002401 Chr.19: 41095137-41095137 on Build GRCh38
rs150922932 Chr.19: 41094989-41094989 on Build GRCh38
rs771376706 Chr.19: 41095146-41095146 on Build GRCh38
rs113962713 Chr.19: 41095006-41095006 on Build GRCh38
rs749481248 Chr.19: 41095140-41095140 on Build GRCh38
rs534584012 Chr.19: 41095057-41095057 on Build GRCh38
rs946422449 Chr.19: 41094994-41094994 on Build GRCh38
rs746175069 Chr.19: 41095148-41095148 on Build GRCh38
rs748438394 Chr.19: 41095045-41095045 on Build GRCh38
rs202218822 Chr.19: 41095020-41095020 on Build GRCh38
rs140896123 Chr.19: 41095002-41095002 on Build GRCh38
rs745865005 Chr.19: 41094963-41094963 on Build GRCh38
rs9916987 Chr.19: 41094972-41094972 on Build GRCh38
rs765414081 Chr.19: 41095022-41095022 on Build GRCh38
rs377513807 Chr.19: 41094962-41094962 on Build GRCh38
rs781120207 Chr.19: 41095131-41095131 on Build GRCh38
rs538929383 Chr.19: 41095053-41095053 on Build GRCh38
rs71579372 Chr.19: 41095042-41095042 on Build GRCh38
rs368488873 Chr.19: 41095147-41095147 on Build GRCh38
rs768826753 Chr.19: 41095030-41095030 on Build GRCh38
rs780068442 Chr.19: 41095025-41095025 on Build GRCh38
rs772139548 Chr.19: 41094964-41094964 on Build GRCh38
rs577832324 Chr.19: 41095023-41095023 on Build GRCh38
rs761348304 Chr.19: 41095070-41095070 on Build GRCh38
rs751569277 Chr.19: 41095100-41095100 on Build GRCh38
rs770204411 Chr.19: 41094998-41094998 on Build GRCh38
rs1015660644 Chr.19: 41095161-41095161 on Build GRCh38
rs199563596 Chr.19: 41094952-41094952 on Build GRCh38
rs554071462 Chr.19: 41095060-41095060 on Build GRCh38
rs757159545 Chr.19: 41094961-41094961 on Build GRCh38
rs753602220 Chr.19: 41095018-41095018 on Build GRCh38
rs770011815 Chr.19: 41095049-41095049 on Build GRCh38
rs774308030 Chr.19: 41095022-41095023 on Build GRCh38
rs755523029 Chr.19: 41094954-41094954 on Build GRCh38
rs768573834 Chr.19: 41094985-41094985 on Build GRCh38

View Full Product Details