Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232760413-232760666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708131 c.417T>G p.S139R Chr.2: 232760517-232760517 on Build GRCh38
1017858 c.413G>T p.R138I Chr.2: 232760513-232760513 on Build GRCh38
1661976 c.411A>T p.Q137H Chr.2: 232760511-232760511 on Build GRCh38
4764823 c.446G>A p.R149Q Chr.2: 232760546-232760546 on Build GRCh38
1017859 c.474G>A p.S158S Chr.2: 232760574-232760574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755653896 Chr.2: 232760434-232760434 on Build GRCh38
rs796163453 Chr.2: 232760503-232760503 on Build GRCh38
rs368804161 Chr.2: 232760574-232760574 on Build GRCh38
rs149250246 Chr.2: 232760485-232760485 on Build GRCh38
rs765820402 Chr.2: 232760629-232760629 on Build GRCh38
rs745679441 Chr.2: 232760454-232760455 on Build GRCh38
rs140796133 Chr.2: 232760475-232760475 on Build GRCh38
rs372442274 Chr.2: 232760596-232760596 on Build GRCh38
rs775586502 Chr.2: 232760597-232760597 on Build GRCh38
rs545447550 Chr.2: 232760573-232760573 on Build GRCh38
rs868580254 Chr.2: 232760579-232760579 on Build GRCh38
rs1043369109 Chr.2: 232760435-232760435 on Build GRCh38
rs778640981 Chr.2: 232760548-232760548 on Build GRCh38
rs767317038 Chr.2: 232760609-232760609 on Build GRCh38
rs114255595 Chr.2: 232760642-232760642 on Build GRCh38
rs770696747 Chr.2: 232760445-232760448 on Build GRCh38
rs746052538 Chr.2: 232760584-232760584 on Build GRCh38
rs778615767 Chr.2: 232760445-232760446 on Build GRCh38
rs372364086 Chr.2: 232760454-232760454 on Build GRCh38
rs763370195 Chr.2: 232760601-232760601 on Build GRCh38
rs1041270281 Chr.2: 232760451-232760451 on Build GRCh38
rs575759510 Chr.2: 232760615-232760615 on Build GRCh38
rs747926777 Chr.2: 232760563-232760563 on Build GRCh38
rs754878774 Chr.2: 232760540-232760540 on Build GRCh38
rs762243537 Chr.2: 232760607-232760607 on Build GRCh38
rs774910743 Chr.2: 232760605-232760605 on Build GRCh38
rs745522498 Chr.2: 232760555-232760555 on Build GRCh38
rs1013641436 Chr.2: 232760632-232760632 on Build GRCh38
rs375429302 Chr.2: 232760603-232760603 on Build GRCh38
rs374364876 Chr.2: 232760477-232760477 on Build GRCh38
rs114353204 Chr.2: 232760620-232760620 on Build GRCh38

View Full Product Details