Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61720668-61720939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAGLA
Entrez Gene ID
747
Gene Name
diacylglycerol lipase alpha
Gene Aliases
C11orf11, DAGL(ALPHA), DAGLALPHA, NSDDR
UniGene
Hs.241564
Gene Chromosome Location
Chr.11: 61680184-61747002 on Build GRCh38
Transcripts
NM_006133.2 XM_017018238.1 XM_017018239.1 XM_017018240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564621 c.129_130GG>AA p.V44I Chr.11: 61720712-61720713 on Build GRCh38
6133841 c.272G>T p.R91L Chr.11: 61720855-61720855 on Build GRCh38
4622209 c.250G>T p.G84C Chr.11: 61720833-61720833 on Build GRCh38
4165870 c.306G>A p.L102L Chr.11: 61720889-61720889 on Build GRCh38
3450737 c.271C>T p.R91C Chr.11: 61720854-61720854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752345548 Chr.11: 61720761-61720761 on Build GRCh38
rs770905349 Chr.11: 61720703-61720703 on Build GRCh38
rs917067468 Chr.11: 61720748-61720748 on Build GRCh38
rs553742435 Chr.11: 61720868-61720868 on Build GRCh38
rs774912823 Chr.11: 61720732-61720732 on Build GRCh38
rs924308928 Chr.11: 61720830-61720830 on Build GRCh38
rs756062957 Chr.11: 61720912-61720912 on Build GRCh38
rs754533394 Chr.11: 61720758-61720758 on Build GRCh38
rs773906365 Chr.11: 61720694-61720694 on Build GRCh38
rs752257511 Chr.11: 61720881-61720881 on Build GRCh38
rs140212906 Chr.11: 61720871-61720871 on Build GRCh38
rs758107539 Chr.11: 61720762-61720762 on Build GRCh38
rs757105086 Chr.11: 61720766-61720766 on Build GRCh38
rs777615529 Chr.11: 61720695-61720695 on Build GRCh38
rs774822524 Chr.11: 61720847-61720847 on Build GRCh38
rs780346829 Chr.11: 61720797-61720797 on Build GRCh38
rs769921060 Chr.11: 61720796-61720796 on Build GRCh38
rs147759229 Chr.11: 61720727-61720727 on Build GRCh38
rs751307974 Chr.11: 61720903-61720903 on Build GRCh38
rs991745197 Chr.11: 61720746-61720746 on Build GRCh38
rs762384427 Chr.11: 61720733-61720733 on Build GRCh38
rs781212877 Chr.11: 61720775-61720775 on Build GRCh38
rs753243808 Chr.11: 61720749-61720749 on Build GRCh38
rs763341853 Chr.11: 61720731-61720731 on Build GRCh38
rs750369177 Chr.11: 61720911-61720911 on Build GRCh38
rs972629226 Chr.11: 61720706-61720706 on Build GRCh38
rs200113525 Chr.11: 61720724-61720724 on Build GRCh38
rs201994401 Chr.11: 61720829-61720829 on Build GRCh38
rs757090449 Chr.11: 61720905-61720905 on Build GRCh38
rs762296637 Chr.11: 61720849-61720849 on Build GRCh38
rs750140845 Chr.11: 61720906-61720906 on Build GRCh38
rs796499509 Chr.11: 61720877-61720877 on Build GRCh38
rs764732794 Chr.11: 61720759-61720759 on Build GRCh38
rs745876087 Chr.11: 61720787-61720787 on Build GRCh38
rs999757225 Chr.11: 61720872-61720872 on Build GRCh38
rs373356660 Chr.11: 61720764-61720764 on Build GRCh38
rs1054086042 Chr.11: 61720880-61720880 on Build GRCh38
rs770521112 Chr.11: 61720823-61720823 on Build GRCh38
rs984036581 Chr.11: 61720708-61720708 on Build GRCh38
rs777523166 Chr.11: 61720763-61720763 on Build GRCh38
rs762563063 Chr.11: 61720885-61720885 on Build GRCh38
rs746939894 Chr.11: 61720700-61720700 on Build GRCh38
rs780923357 Chr.11: 61720908-61720908 on Build GRCh38
rs749507427 Chr.11: 61720828-61720828 on Build GRCh38
rs763768642 Chr.11: 61720897-61720897 on Build GRCh38
rs772407641 Chr.11: 61720856-61720856 on Build GRCh38
rs780258755 Chr.11: 61720920-61720920 on Build GRCh38
rs768105248 Chr.11: 61720743-61720743 on Build GRCh38
rs776542702 Chr.11: 61720707-61720707 on Build GRCh38

View Full Product Details