Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 12995012-12995239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC2
Entrez Gene ID
60528
Gene Name
elaC ribonuclease Z 2
Gene Aliases
COXPD17, ELC2, HPC2
UniGene
Hs.434232
Gene Chromosome Location
Chr.17: 12991612-13018064 on Build GRCh38
Transcripts
NM_001165962.1 NM_018127.6 NM_173717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3514403 c.1816C>T p.P606S Chr.17: 12995055-12995055 on Build GRCh38
3514401 c.1856C>T p.P619L Chr.17: 12995015-12995015 on Build GRCh38
3514402 c.1850C>T p.S617F Chr.17: 12995021-12995021 on Build GRCh38
4634155 c.1817C>A p.P606H Chr.17: 12995054-12995054 on Build GRCh38
3937257 c.1810A>G p.M604V Chr.17: 12995061-12995061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776612685 Chr.17: 12995108-12995108 on Build GRCh38
rs756645244 Chr.17: 12995040-12995040 on Build GRCh38
rs556412683 Chr.17: 12995192-12995192 on Build GRCh38
rs752421143 Chr.17: 12995201-12995201 on Build GRCh38
rs776246924 Chr.17: 12995142-12995142 on Build GRCh38
rs368043147 Chr.17: 12995076-12995076 on Build GRCh38
rs772002636 Chr.17: 12995094-12995094 on Build GRCh38
rs981112913 Chr.17: 12995059-12995059 on Build GRCh38
rs528083847 Chr.17: 12995176-12995176 on Build GRCh38
rs114943735 Chr.17: 12995212-12995212 on Build GRCh38
rs758757012 Chr.17: 12995205-12995205 on Build GRCh38
rs73981604 Chr.17: 12995137-12995137 on Build GRCh38
rs749375920 Chr.17: 12995063-12995063 on Build GRCh38
rs759921340 Chr.17: 12995166-12995166 on Build GRCh38
rs566410184 Chr.17: 12995200-12995200 on Build GRCh38
rs368138149 Chr.17: 12995098-12995098 on Build GRCh38
rs571928216 Chr.17: 12995204-12995204 on Build GRCh38
rs530546046 Chr.17: 12995122-12995122 on Build GRCh38
rs1004609952 Chr.17: 12995162-12995162 on Build GRCh38
rs746016313 Chr.17: 12995083-12995083 on Build GRCh38
rs913207452 Chr.17: 12995051-12995051 on Build GRCh38
rs1041978587 Chr.17: 12995172-12995172 on Build GRCh38
rs1043062927 Chr.17: 12995102-12995102 on Build GRCh38
rs570984569 Chr.17: 12995129-12995129 on Build GRCh38
rs1055698524 Chr.17: 12995133-12995133 on Build GRCh38
rs951245217 Chr.17: 12995039-12995039 on Build GRCh38
rs545514750 Chr.17: 12995135-12995135 on Build GRCh38
rs753264388 Chr.17: 12995034-12995034 on Build GRCh38
rs778310432 Chr.17: 12995043-12995043 on Build GRCh38
rs753204082 Chr.17: 12995073-12995074 on Build GRCh38
rs148056623 Chr.17: 12995105-12995105 on Build GRCh38
rs199813028 Chr.17: 12995099-12995099 on Build GRCh38

View Full Product Details