Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36098257-36098512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A2
Entrez Gene ID
1296
Gene Name
collagen type VIII alpha 2
Gene Aliases
FECD, FECD1, PPCD, PPCD2
UniGene
Hs.353001
Gene Chromosome Location
Chr.1: 36095236-36126207 on Build GRCh38
Transcripts
NM_001294347.1 NM_005202.3 XM_005270477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402750 c.1343G>T p.G448V Chr.1: 36098338-36098338 on Build GRCh38
4515797 c.1317_1318GG>AA p.V440M Chr.1: 36098363-36098364 on Build GRCh38
4962952 c.1209G>A p.G403G Chr.1: 36098472-36098472 on Build GRCh38
3985013 c.1176T>C p.I392I Chr.1: 36098505-36098505 on Build GRCh38
6009216 c.1240C>T p.P414S Chr.1: 36098441-36098441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890352210 Chr.1: 36098301-36098302 on Build GRCh38
rs187303432 Chr.1: 36098430-36098430 on Build GRCh38
rs758598302 Chr.1: 36098443-36098443 on Build GRCh38
rs910331711 Chr.1: 36098355-36098355 on Build GRCh38
rs747018538 Chr.1: 36098464-36098464 on Build GRCh38
rs775601050 Chr.1: 36098346-36098346 on Build GRCh38
rs867635771 Chr.1: 36098421-36098421 on Build GRCh38
rs921721215 Chr.1: 36098276-36098276 on Build GRCh38
rs200364502 Chr.1: 36098288-36098288 on Build GRCh38
rs768923775 Chr.1: 36098481-36098481 on Build GRCh38
rs867250639 Chr.1: 36098399-36098399 on Build GRCh38
rs727504229 Chr.1: 36098317-36098318 on Build GRCh38
rs370063465 Chr.1: 36098381-36098381 on Build GRCh38
rs553809063 Chr.1: 36098426-36098426 on Build GRCh38
rs769226465 Chr.1: 36098354-36098354 on Build GRCh38
rs1035673350 Chr.1: 36098387-36098387 on Build GRCh38
rs772297339 Chr.1: 36098423-36098423 on Build GRCh38
rs571229508 Chr.1: 36098363-36098363 on Build GRCh38
rs751018601 Chr.1: 36098394-36098394 on Build GRCh38
rs745587752 Chr.1: 36098433-36098433 on Build GRCh38
rs762703964 Chr.1: 36098385-36098385 on Build GRCh38
rs778992404 Chr.1: 36098283-36098283 on Build GRCh38
rs201235688 Chr.1: 36098380-36098380 on Build GRCh38
rs1048444640 Chr.1: 36098379-36098379 on Build GRCh38
rs1057299696 Chr.1: 36098416-36098416 on Build GRCh38
rs752690999 Chr.1: 36098405-36098405 on Build GRCh38
rs866394027 Chr.1: 36098480-36098480 on Build GRCh38
rs867019752 Chr.1: 36098459-36098459 on Build GRCh38
rs182708720 Chr.1: 36098358-36098358 on Build GRCh38
rs887324561 Chr.1: 36098400-36098400 on Build GRCh38
rs762261875 Chr.1: 36098375-36098375 on Build GRCh38
rs80358192 Chr.1: 36098332-36098332 on Build GRCh38
rs767441504 Chr.1: 36098396-36098396 on Build GRCh38
rs765975038 Chr.1: 36098386-36098386 on Build GRCh38
rs747108574 Chr.1: 36098353-36098353 on Build GRCh38
rs780153991 Chr.1: 36098458-36098458 on Build GRCh38
rs80358191 Chr.1: 36098318-36098318 on Build GRCh38
rs977324392 Chr.1: 36098295-36098295 on Build GRCh38
rs1010496595 Chr.1: 36098422-36098422 on Build GRCh38
rs754492511 Chr.1: 36098395-36098395 on Build GRCh38

View Full Product Details