Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41482365-41482626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN2
Entrez Gene ID
1907
Gene Name
endothelin 2
Gene Aliases
ET-2, ET2, PPET2
UniGene
Hs.1407
Gene Chromosome Location
Chr.1: 41478775-41484699 on Build GRCh38
Transcripts
NM_001302269.1 NM_001956.4 NR_126098.1 XM_017000512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404774 c.244G>T p.G82* Chr.1: 41482566-41482566 on Build GRCh38
3489654 c.281G>C p.R94T Chr.1: 41482529-41482529 on Build GRCh38
7365856 c.268C>T p.R90C Chr.1: 41482542-41482542 on Build GRCh38
426274 c.263G>C p.R88P Chr.1: 41482547-41482547 on Build GRCh38
6355720 c.260G>A p.R87H Chr.1: 41482550-41482550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761370959 Chr.1: 41482537-41482537 on Build GRCh38
rs918182946 Chr.1: 41482432-41482432 on Build GRCh38
rs756588267 Chr.1: 41482493-41482493 on Build GRCh38
rs144539806 Chr.1: 41482542-41482542 on Build GRCh38
rs748626790 Chr.1: 41482489-41482489 on Build GRCh38
rs540846506 Chr.1: 41482502-41482502 on Build GRCh38
rs767394635 Chr.1: 41482437-41482437 on Build GRCh38
rs984881184 Chr.1: 41482396-41482396 on Build GRCh38
rs373903543 Chr.1: 41482603-41482603 on Build GRCh38
rs147049205 Chr.1: 41482483-41482483 on Build GRCh38
rs760617539 Chr.1: 41482456-41482456 on Build GRCh38
rs746112557 Chr.1: 41482557-41482557 on Build GRCh38
rs757449064 Chr.1: 41482550-41482550 on Build GRCh38
rs1026774838 Chr.1: 41482554-41482554 on Build GRCh38
rs201795232 Chr.1: 41482558-41482558 on Build GRCh38
rs768322245 Chr.1: 41482560-41482560 on Build GRCh38
rs778406208 Chr.1: 41482549-41482549 on Build GRCh38
rs996084776 Chr.1: 41482509-41482509 on Build GRCh38
rs772020788 Chr.1: 41482508-41482508 on Build GRCh38
rs763531751 Chr.1: 41482460-41482460 on Build GRCh38
rs764747986 Chr.1: 41482573-41482573 on Build GRCh38
rs11572355 Chr.1: 41482419-41482419 on Build GRCh38
rs764024548 Chr.1: 41482518-41482518 on Build GRCh38
rs994669973 Chr.1: 41482417-41482417 on Build GRCh38
rs775304398 Chr.1: 41482510-41482510 on Build GRCh38
rs755376927 Chr.1: 41482486-41482486 on Build GRCh38
rs757998837 Chr.1: 41482544-41482544 on Build GRCh38
rs759466563 Chr.1: 41482434-41482434 on Build GRCh38
rs200513885 Chr.1: 41482548-41482548 on Build GRCh38
rs376618881 Chr.1: 41482545-41482545 on Build GRCh38
rs751155668 Chr.1: 41482597-41482597 on Build GRCh38
rs558455490 Chr.1: 41482475-41482475 on Build GRCh38
rs1056702351 Chr.1: 41482426-41482426 on Build GRCh38
rs779091830 Chr.1: 41482551-41482551 on Build GRCh38
rs138778457 Chr.1: 41482534-41482534 on Build GRCh38
rs762076357 Chr.1: 41482534-41482535 on Build GRCh38
rs543002523 Chr.1: 41482541-41482541 on Build GRCh38
rs112514293 Chr.1: 41482595-41482595 on Build GRCh38
rs760540746 Chr.1: 41482512-41482512 on Build GRCh38
rs1049539130 Chr.1: 41482532-41482532 on Build GRCh38
rs766119457 Chr.1: 41482596-41482596 on Build GRCh38
rs781498204 Chr.1: 41482487-41482487 on Build GRCh38
rs71648549 Chr.1: 41482570-41482570 on Build GRCh38
rs200003277 Chr.1: 41482583-41482583 on Build GRCh38
rs201905094 Chr.1: 41482497-41482497 on Build GRCh38
rs761698280 Chr.1: 41482565-41482565 on Build GRCh38
rs752612340 Chr.1: 41482453-41482453 on Build GRCh38
rs538985161 Chr.1: 41482547-41482547 on Build GRCh38
rs776678882 Chr.1: 41482562-41482562 on Build GRCh38
rs967704945 Chr.1: 41482527-41482527 on Build GRCh38
rs373317935 Chr.1: 41482420-41482420 on Build GRCh38
rs749454994 Chr.1: 41482498-41482498 on Build GRCh38
rs746888124 Chr.1: 41482559-41482559 on Build GRCh38
rs554208821 Chr.1: 41482505-41482505 on Build GRCh38
rs538962790 Chr.1: 41482402-41482402 on Build GRCh38
rs753329833 Chr.1: 41482471-41482471 on Build GRCh38

View Full Product Details